Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CN0372S06

Millipore

Polygard® CN Cartridge Filter

nominal pore size 0.3 μm, cartridge nominal length × diam. 20 in. (564.0-572.0 mm) × 2.75 in. (70 mm)

Sign Into View Organizational & Contract Pricing

Select a Size

6 PKG
$2,693.00

$2,693.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
6 PKG
$2,693.00

About This Item

UNSPSC Code:
23151806

$2,693.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.

material

polypropylene filter

Quality Level

feature

hydrophilic

manufacturer/tradename

Polygard

parameter

4.8 bar max. differential pressure (70 psid) at 20 °C

cartridge nominal length × diam.

20 in. (564.0-572.0 mm) × 2.75 in. (70 mm)

impurities

≤0.25 EU/mL bacterial endotoxins (LAL test, aqueous extraction)

gravimetric extractables

≤10 mg

pore size

0.3 μm nominal pore size
0.3 μm pore size

input

sample type liquid

cartridge code

Code 7 (2-226; O-Ring with locking tab and spear)

Looking for similar products? Visit Product Comparison Guide

General description

Polygard® CN products are nominally rated filters designed for particle removal applications in liquids and gases. The pleated structure of Polygard® CN filter media minimizes pressure differentials during the filtration process, and the all-polypropylene construction of these filters conveys low levels of extractables and broad chemical compatibility. Downstream filter performance is enhanced due to efficient particle and microorganism reduction.

Preparation Note

Sterilization: May be autoclaved for 10 cycles of 30 minutes at 126 °C or steam sterilized for 10 cycles for 30 minutes at 126 °C or hot water sanitized at 80 °C maximum for 30 minutes.

Analysis Note

Gravimetric Extractables: After autoclaving and a 24-hour soak in ASTM® Type 1 reagent grade water at controlled room temperature, meets the USP Oxidizable Substances Test requirements of <50 mg.

Other Notes

Directions for Use:

  • Organism Retention: Microorganism reduction
  • Mode of Action: Filtration (nominal)
  • Application: BioProcessing
  • Intended Use: Efficient particle and microorganism reduction
  • Instructions for Use: Please refer to the Polygard® and Clarigard® Cartridges user guide
  • Storage Statement: Store in dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
CLARIGARD is a registered trademark of Merck KGaA, Darmstadt, Germany
POLYGARD is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service