Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CC126

Sigma-Aldrich

Caspase 1, recombinant human active

Synonym(s):

Recombinant caspase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.41

biological source

human

recombinant

expressed in E. coli

form

solid

manufacturer/tradename

Chemicon®

technique(s)

activity assay: suitable

UniProt accession no.

shipped in

dry ice

General description

Caspase-1 (also known as ICE, Interleukin-1beta-Converting Enzyme) is the prototypical member of the caspase family of cysteine proteases. Caspase-1 exists in cells as an inactive 45 kDa pro-enzyme. The pro-enzyme is matured by proteolysis to yield large (20 kD) and small (10 kD) subunits. The active enzyme is a heterotetramer consisting of two large and two small subunits. To date the mechanism of regulation of caspase-1 activation is complex and poorly understood. In THP-1 cells, a large proportion of the caspase-1 is present in the inactive pro-enzyme form.

The active recombinant human caspase-1 was expressed in E. coli. The expressed caspase-1 spontaneously undergoes autoprocessing to yield the subunits characteristic of the native enzyme. The recombinant caspase-1 preferentially cleaves the substrates consisting of consensus sequence YVAD (e.g., YVAD-AFC and YVAD-pNA). The recombinant enzyme can also be used as a positive control in caspase assays or in determining the specificity of caspase substrates.

Recommended usage: 0.5-1 unit for fluorometric caspase assays and 2-4 units for colorimetric caspase assays. Optimal working dilutions must be determined by end user.

Physical form

Lyophilized. Reconstitute in 25 μL PBS containing 15% glycerol.

Storage and Stability

Maintain unopened and rehydrated product at -70°C in undiluted, freeze-thaw protected, tightly sealed aliquots. Avoid repeated freeze/thaw cycles. Product stable for 6 months from date of purchased if handled appropriately.

Analysis Note

Partially purified recombinant caspase 3 with a full length HIS-6-tag. Approximately 10-20% caspase 1 protein. The remainder is E.Coli proteins. Approximately 0.8-2 mg total protein/U of activity; E.Coli proteins have no reactivity.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qianyue Chen et al.
European journal of immunology, 51(5), 1234-1245 (2021-01-18)
Pyroptosis is a type of acute cell death that mainly occurs in immune cells. It is characterized with robust release of inflammatory cytokines and has emerged to play a critical role in the pathogenesis of sepsis-associated immune disorders. In this
Timothy Connelley et al.
Veterinary research, 42, 37-37 (2011-02-25)
Natural killer (NK) cells are well recognized as playing a key role in innate immune defence through cytokine production and cytotoxic activity; additionally recent studies have identified several novel NK cell functions. The ability to study NK cells in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service