Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BX1634

Supelco

Buffer pH 4.0 Clear

Synonym(s):

buffer solutions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

analytical standard

Quality Level

agency

traceable to NIST

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

shipped in

ambient

storage temp.

15-25°C

Application

This pH 4.0 buffer solution can be used in routine analytical processes for the calibration of pH meters and for maintaining pH of solutions.

Features and Benefits

  • A ready-to-use solution available in a sealed bottle with convenient packaging to protect against contamination
  • Traceable to NIST standards
  • Accompanied with a certificate of analysis (CoA) for an effective audit
  • Accurate up to +/- 0.01 at 25 °C
  • The expiry date of the product can be found in the CoA
  • Uncertainty value calculated includes for the duration of transportation and storage as well

Specifications

Tolerance: ±0.01

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service