BVLMC0120
Beviflex liquid media cassette
For bioburden and water analysis with the Beviflex™ system
Synonym(s):
Media cassette
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116129
Recommended Products
material
clear device
polypropylene
sterility
sterile; ethylene oxide treated
manufacturer/tradename
Beviflex™
input
sample type beverage(s)
application(s)
bioburden testing
food and beverages
shipped in
ambient
General description
Beviflex™ Liquid Media Cassette is designed to be used with Beviflex™ system for convenient, rapid, and reliable bioburden testing. The liquid media cassette helps to streamline the microbial contamination testing workflow with quality assurance and avoiding risks of exogenous contamination. Each plate can hold approximately 2mL of a liquid media dispensed by the user. Broth media in ampoule is typically used in combination with the Beviflex™ Liquid Media Cassette.
Application
Beviflex™ Liquid Media Cassette is used in Bioburden testing and general microbiological analysis of water. It is widely used in the cosmetic and brewing industry.
Legal Information
BEVIFLEX is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service