Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BI0354

Sigma-Aldrich

Capping Reagent B , 15% NMI in Acetonitrile

Biosynthesis Novabiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

20 L
$1,150.00

$1,150.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 L
$1,150.00

About This Item

UNSPSC Code:
41116006

$1,150.00


Check Cart for Availability

Request a Bulk Order

Quality Level

product line

Novabiochem®

form

liquid

reaction suitability

reaction type: Acetylations
reagent type: derivatization reagent

transition temp

flash point 6.0 °C

shipped in

ambient

storage temp.

2-30°C

Application

Biosynthesis Novabiochem®

Analysis Note

Assay 1-Methylimidazole: 13-17%
Assay Acetonitrile: 83-87%
Color (APHA):
Form: Clear liquid
Infrared Spectrum: conforms
Water (H2O): 0-150ppm

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

42.8 °F - closed cup

flash_point_c

6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service