Key Documents
AX5/41-E2-062
NovAseptic® Drive Unit,GMP500DU 0.25kW 7:1 E2 II 1/2 G Ex h
About This Item
UNSPSC Code:
23151800
Pricing and availability is not currently available.
AC/DC input
230 V / 400 V AC, 50 Hz
parameter
atmospheric pressure pressure range
0-40 °C temp. range
weight
18 kg (39.7 lb)
Application
Maximum operating speed has to be validated for each process individually, depending on volume, viscosity etc. The assembled NovAseptic® Mixer may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Packaging
The Drive Unit is packaged in a closed box.
Legal Information
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany