Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AX1699M

Supelco

Methanol

anhydrous AQUASTAR®

Synonym(s):

Methyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OH
CAS Number:
Molecular Weight:
32.04
Beilstein/REAXYS Number:
1098229
MDL number:
UNSPSC Code:
41116121
EC Index Number:
200-659-6

vapor density

1.11 (vs air)

Quality Level

vapor pressure

128 hPa ( 20 °C)
410 mmHg ( 50 °C)
97.68 mmHg ( 20 °C)

assay

≥99.8% (GC)

autoignition temp.

725 °F

expl. lim.

36 %

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.329 (lit.)

bp

64.7 °C (lit.)

mp

−98 °C (lit.)

transition temp

flash point 9.7 °C

density

0.791 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

CO

InChI

1S/CH4O/c1-2/h2H,1H3

InChI key

OKKJLVBELUTLKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

AX1699M-6 and AX1699M-3 are packaged in 38/430 glass bottles.
anhydrous AQUASTAR®

Analysis Note

AppearanceClearAssay (GC)99.8% minCarbonyl (acetone,formaldehyde,acetaldehyde)0.001% maxColor (APHA)10 maxEvaporation residue0.001% maxSolubility in waterTo pass testSubstances darkened by sulfuric acidTo pass testSubstances reducing permanganateTo pass testTitrable acid0.0003meq/g maxTitrable base0.0002meq/g maxWater (H2O)0.01% max
Evaporation Number: 1.9

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service