Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AX1699EN

Supelco

Solvent KN

Synonym(s):

Karl Fischer

Sign Into View Organizational & Contract Pricing


About This Item

form

liquid

Quality Level

transition temp

flash point 39 °C

Storage temp.

room temp

Application

AQUASTAR®
AX1699EN-6 and AX1699EN-5 are packaged in 38/430 glass bottles

Analysis Note

Color:Colorless
Form:Clear liquid
Performance:To pass test
Water (H2O):0.06-0.07%

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B - STOT RE 2 - STOT SE 1

target_organs

Immune system, thymus

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

102.2 °F

flash_point_c

39 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service