Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AX1698D

Supelco

Composite 5K

Synonym(s):

Karl Fischer

Sign Into View Organizational & Contract Pricing


About This Item

form

liquid

transition temp

flash point 42.2 °C

storage temp.

room temp

application

AX1698D-6 and AX1698D-5 are packaged in 38/430 poly coated glass bottles.
Original Formulation AQUASTAR®

Analysis Note

Color:Red-brown
Form:Liquid
Performance:To pass test
Titer (mg water per mL):5.0mg/mL min

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - STOT RE 1 Oral - STOT RE 2 - STOT SE 1

target_organs

Immune system, thymus, Thyroid

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

108.0 °F

flash_point_c

42.2 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service