Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AX0735

Supelco

Aluminum Sulfate, Octadecahydrate

Meets ACS Specifications GR ACS

Synonym(s):

Aluminum Sulfate, Octadecahydrate, Cake alum,Patent alum, Alum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2(SO4)3 · 14-18H2O
CAS Number:
UNSPSC Code:
12352302
EC Index Number:
233-135-0
assay:
98.0-102.0% (complexometric)
grade:
ACS reagent
form:
crystals
solubility:
364 g/L

grade

ACS reagent

Quality Level

assay

98.0-102.0% (complexometric)

form

crystals

impurities

≤0.01% Insoluble matter

color

white

pH

2.5-4.0 (20 °C, 20 g/L in H2O)

mp

90 °C (decomposition)

solubility

364 g/L

anion traces

chloride (Cl-): ≤0.005%

cation traces

Ca: ≤0.01%
Fe: ≤0.002%
K: ≤0.005%
Mg: ≤0.002%
Na: ≤0.02%
heavy metals (as Pb): ≤0.001%

shipped in

ambient

storage temp.

room temp

InChI

1S/2Al.3H2O4S.9H2O/c;;3*1-5(2,3)4;;;;;;;;;/h;;3*(H2,1,2,3,4);9*1H2/q2*+3;;;;;;;;;;;;/p-6

InChI key

KFCWRPFNNIEZRY-UHFFFAOYSA-H

application

Meets ACS Specifications GR ACS

Analysis Note

Assay (complexometric): 98.0-102.0%
Calcium (Ca): 0.01% max
Chloride (Cl): 0.005% max
Color: White
Form: Crystals
Heavy metals (as Pb): 0.001% max
Insoluble matter: 0.01% max
Iron (Fe): 0.002% max
Magnesium (Mg): 0.002% max
Potassium (K): 0.005% max
Sodium (Na): 0.02% max

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service