Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AX0120T

Acetone

Synonym(s):

2-Propanone, Dimethyl Ketone

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

vapor pressure

233 hPa ( 20 °C)

form

liquid

pH

5-6 (20 °C, 395 g/L in H2O)

bp

56.2 °C/1013 hPa

mp

-95.4 °C

transition temp

flash point <-20 °C

format

neat

storage temp.

room temp

Application

AX0120P-4 is packaged in a poly-coated glass bottle
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Aldehyde (as HCHO):0.002% max
Assay (GC, corrected for water content):99.5% min
Color (APHA):10 max
Evaporation residue:0.001% max
Form:Clear liquid
Isopropyl alcohol:0.05% max
Methanol:0.05% max
Solubility in water:To pass test
Substances reducing permanganate:To pass test
Titrable acid:0.0003meq/g max
Titrable base:0.0006meq/g max
Water (H2O):0.5% max

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service