Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATBPRNT11

Millipore

Printer Expendables for printer MXPPPRN01 (5 rolls; 2 ink cartridges)

The Printer Expendables are an accessory for the Milliflex Plus Pump. It is a refill pack for the Milliflex Plus Printer.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.11

input

sample type pharmaceutical(s)

application(s)

environmental

compatibility

for use with Milliflex® Plus Printer

shipped in

ambient

Components

5 rolls of paper

2 ink cartridges

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service