Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ATBCSEV01

Millipore

M Air T Sieve for Cone Adapter

The M Air T sieve for cone adapter is uniquely micro-perforated and allows sampling of up to 1 cubic meter of air.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
eCl@ss:
25140604
NACRES:
NB.11

manufacturer/tradename

M Air T

input

sample type pharmaceutical(s)

application(s)

environmental

compatibility

for use with M Air T Compressed Gas and Air Test Kit

shipped in

ambient

Legal Information

M AIR T is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service