Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AP202C

Sigma-Aldrich

Goat Anti-Rat light chain Antibody, Cy3 conjugate

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
$260.00

$260.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
0.5 MG
$260.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

$260.00


Check Cart for Availability

Request a Bulk Order

biological source

goat

Quality Level

conjugate

CY3 conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

General description

Light chains covalently bound to a pair of heavy chains to form an antibody molecule. Immunoglobulin light chains occur in two types, kappa or lambda. Light chain′s mass is about 25,000 Daltons per mole. Each light chain consists of about 250 amino-acids long. These are usually designated by the Greek letters kappa and lambda. The ratio of kappa to lambda found in the immunoglobulin population varies by species.

Specificity

Minimal cross-reaction with bovine, goat, horse, human, mouse, rabbit, and sheep.

Application

Detect light chain using this Goat anti-Rat light chain Antibody, Cy3 conjugate validated for use in ELISA & WB.
Western Blotting: 1:100 -1:800

ELISA: 1:100 -1:800

Optimal working dilutions must be determined by the end user.

Target description

25 kDa

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service