Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AP10047E0

Millipore

Absorbent pads sterile EO sterilized 47 mm, 10 envelopes of 10 pads

sterile absorbent pads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122101
eCl@ss:
32030209

material

cellulose pad(s)
clear device
polystyrene

sterility

sterile; ethylene oxide treated

diam.

47 mm

input

sample type cider
sample type beer
sample(s) (environmental)
sample type sport drink
sample type beverage(s)
sample type cosmetics
sample type food(s)
sample type soft drink
sample type bottled water
sample type wine

application(s)

bioburden testing
clinical
cosmetics
environmental
food and beverages
industrial qc
pharmaceutical

shipped in

ambient

Application

Dispensing of sterile absorbent pads for microbiological analysis of beverages.

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service