Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ACS500302

Millipore

Amicon® Pro Purification System with Ultra-0.5 Device

Amicon Pro Purification System - A 3 kDa Centrifugal Tool collapsing protein purifcation with concentraion & buffer exchange.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
eCl@ss:
36100101
NACRES:
NB.24

material

Amicon Ultra styrene-butadiene device
Ultracel® regenerated cellulose membrane (Amicon Ultra-0.5; low binding)
polyethylene cap
polyethylene frit
polypropylene device (for exchange)
polypropylene frit
polypropylene tube (AU0.5; for collection)
polypropylene tube (for collection)
polypropylene tube (holder)

sterility

non-sterile

product line

Amicon®

manufacturer/tradename

Amicon® Pro

parameter

10 mL sample volume

technique(s)

buffer exchange: suitable
protein purification: suitable (antibody labeling)
protein purification: suitable (concentration)
protein purification: suitable (enrichment)

L

12 cm (4.7 in.)

diam.

2.97 cm

filtration area

1 cm2

working volume

10 mL

pore size

3 kDa NMWCO

shipped in

ambient

General description

The Amicon® Pro Purification System 2 pack includes: 2 Amicon Ultra NMWCO 3 kDa filters, 2 Exchange devices, and 2 Collection tubes with cap.

Application

Amicon Pro Purification System - A 3 kDa Centrifugal Tool collapsing protein purifcation with concentraion & buffer exchange.

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service