Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABT1486

Sigma-Aldrich

Anti-CD31/PECAM-1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Platelet endothelial cell adhesion molecule, CD31, EndoCAM, GPIIA′, PECA1, PECAM-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PECAM1(5175)

General description

Platelet endothelial cell adhesion molecule (UniProt P16284; also known as CD31, EndoCAM, GPIIA′, PECA1, PECAM-1) is encoded by the PECAM1 gene (Gene ID 5175) in human. PECAM-1 (CD31) is a 130-kD cell adhesion molecule that is expressed at the junctions of all continuous endothelia, as well as most bone marrow-derived hematopoietic cells, including platelets, monocytes, granulocytes and some lymphocytes. The human PECAM1 gene produces six spliced isoforms with varying lengths in cytoplasmic domain, with the isoform Long being the major form expressed in human tissues and endothelial cells (ECs), and isoform delta 14-15 being the predominantly expressed murine form. PECAM-1 plays an import role in maintaining vascular integrity and function by mediating ECs interactions with pericytes and smooth muscle cells (SMCs). PECAM-1 is initially produced with a signal peptide (a.a. 1-27) sequence, the removal of which yields the mature protein with a large extracellular region (a.a. 28-601) that contains six Ig-like C2-type domains, followed by a transmembrane seqment (a.a. 602-620) and a cytoplasmic tail (a.a. 621-738 of isoform Long) that plays an essential role in PECAM-1-mediated cellular signaling.

Specificity

Expected to react with all six spliced isoforms of human PECAM-1 reported by UniProt (P16284).

Immunogen

KLH-conjugated linear peptide corresponding to a cytoplasmic domain seqence of human CD31/PECAM-1.

Application

Anti-CD31/PECAM-1 Antibody is an antibody against CD31/PECAM-1. The CD31 antibody is used in Western Blotting and Flow Cytometry applications.
Flow Cytometry Analysis: 0.1 µg of this antibody from a representative lot detected CD31/PECAM-1 in HUVECs and Jurkat cells.

Quality

Evaluated by Western Blotting in HUVECs lysate.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected CD31/PECAM-1 in 10 µg of HUVEC lysate.

Target description

~130 kDa observed. Target band size is larger than the calculated molecular weights due to glycosylation. 79.58 kDa (isoform Long), 77.52 kDa (isoform Delta12), 77.25 kDa (isoform Delta13), 77.46 kDa (isoform Delta14), 77.30 kDa (isoform Delta14-15), 78.42 kDa (isoform Delta15) calculated.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service