Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS2083

Sigma-Aldrich

Anti-TFAM

serum, from rabbit

Synonym(s):

MtTFA, Mitochondrial transcription factor 1, mtTF1, Transcription factor 6, TCF-6, Transcription factor 6-like 2, Transcription factor A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... TFAM(7019)

General description

Transcription factor A, mitochondrial (UniProt: Q00059; also known as MtTFA, Mitochondrial transcription factor 1, mtTF1, Transcription factor 6, TCF-6, Transcription factor 6-like 2, TFAM) is encoded by the TFAM (also known as TCF6, TCF6L2) gene (Gene ID: 7019) in human. TFAM is essential for human mtDNA transcription and acts as a key regulator of mtDNA copy number. It is synthesized as a precursor form with a mitochondrial targeting peptide that is cleaved following import into mitochondria. TFAM consists of an amino-terminal HMG domain, a basic linker region, a second HMG domain and a basic carboxy-terminal tail. TFAM binds to the mitochondrial light strand promoter and functions in mitochondrial transcription regulation. It is required for accurate and efficient promoter recognition by the mitochondrial RNA polymerase. TFAM is reported to promote transcription initiation from the HSP1 and the light strand promoter by binding immediately upstream of transcriptional start sites. It is also able to unwind DNA and bend the mitochondrial light strand promoter DNA into a U-turn shape via its HMG boxes. Tissue-specific depletion of TFAM protein is shown to deplete mitochondrial tDNA and causes severe respiratory chain deficiency. (Ref.: Ekstrand, MI et al. (2004). Hum. Mol. Genet. 13(9): 935-944).

Specificity

This polyclonal antibody specifically detects TFAM in human.

Immunogen

His-Sumo recombinant fusion protein corresponding to 204 amino acids of human TFAM. The signal peptide was removed prior to immunization.

Application

Detect TFAM, mitochondrial using this rabbit polyclonal Anti-TFAM, Cat. No. ABS2083. It has been tested for use in Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A representative lot detected TFAM in transfected Hela cell Lysate (Nouws, J., et. al. (2016). J Biol Chem. 291(2):989-97).

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected TFAM in Huh7.5 cell lysate (Courtesy of Dr. Suresh Sharma, Cameron Laboratory, Pennsylvania State University, University Park, PA).


Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected TFAM in 10 µg of HeLa cell lysate.

Target description

~25 kDa observed; 29.00 kDa calculated.. Uncharacterized bands may be observed in some lysate(s).

Physical form

Rabbit polyclonal antiserum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shizheng Huang et al.
PLoS biology, 16(9), e2005233-e2005233 (2018-09-19)
While Notch signaling has been proposed to play a key role in fibrosis, the direct molecular pathways targeted by Notch signaling and the precise ligand and receptor pair that are responsible for kidney disease remain poorly defined. In this study
Xuan Xu et al.
Cardiovascular diagnosis and therapy, 10(3), 453-469 (2020-07-23)
Myocardial mitochondrial dysfunction is the leading cause of chronic heart failure (CHF). Increased reactive oxygen species (ROS) levels, disruption of mitochondrial biogenesis and mitochondrial Ca2+([Ca2+]m) homeostasis and reduction of the mitochondrial membrane potential (ΔΨm) cause myocardial mitochondrial dysfunction. Therefore, treating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service