Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

ABS186

Sigma-Aldrich

Anti-Spred-1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Sprouty-related, EVH1 domain-containing protein 1, Spred-1, hSpred1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SPRED1(161742)

General description

Spred1 is a tyrosine kinase substrate which has a cysteine-rich domain which resembles the SPR domain at the carboxy terminus. Spred1 associated with Ras, blocks phosphorylation of Raf and disrupts MAP kinase signaling during the differentiation of neuronal cells and myocytes. A mutation in Spred1 has been linked to Lugius syndrome, an autosomal dominant RAS-MAPK pathway syndrome.

Immunogen

KLH-conjugated linear peptide corresponding to human Spred-1.

Application

Anti-Spred-1 Antibody is an antibody against Spred-1 for use in WB.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors

Quality

Evaluated by Western Blot in HEK293 cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Spred-1 on 10 µg of HEK293 cell lysate.

Target description

~50 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiyong Lei et al.
Journal of cellular and molecular medicine, 19(8), 1994-2005 (2015-05-07)
Arteriogenesis is a complicated process induced by increased local shear-and radial wall-stress, leading to an increase in arterial diameter. This process is enhanced by growth factors secreted by both inflammatory and endothelial cells in response to physical stress. Although therapeutic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service