ABS1676
Anti-VPS25
from rabbit
Synonym(s):
Vacuolar protein-sorting-associated protein 25, hVps25, Dermal papilla-derived protein 9, ELL-associated protein of 20 kDa, ESCRT-II complex subunit VPS25
Select a Size
About This Item
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human, monkey
technique(s)
immunofluorescence: suitable
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... VPS25(84313)
General description
Specificity
Immunogen
Application
Western Blotting Analysis: A 1:500 & 1:1,000 dilution from a representative lot detected VPS25 in lysate of transfected HEK293T cells (Sandrin, V., et. al. (2013). Retrovirology. 10:104; Langelier, C., et. al. (2006). J Virol. 80(19):9465-80).
Signaling
Quality
Western Blotting Analysis: A 1:2,000 dilution of this antibody detected VPS25 in 10 µg of HEK293 cell lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service