Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABS1671

Sigma-Aldrich

Anti-Calreticulin Antibody, arginylated (Nt-Glu18)

from rabbit, purified by affinity chromatography

Synonym(s):

Calreticulin, Nt-Glu18 arginylated, Calregulin, Nt-Glu18 arginylated, CRP55, Nt-Glu18 arginylated, CRT, Nt-Glu18 arginylated, Endoplasmic reticulum resident protein 60, Nt-Glu18 arginylated, ERp60, Nt-Glu18 arginylated, grp60, Nt-Glu18 arginylated, HACBP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$444.00

$444.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1681

Request a Bulk Order

Select a Size

Change View
100 μG
$444.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

$444.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1681

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CALR(811)

General description

Calreticulin (UniProt P27797; also known as Calregulin, CRP55, CRT, Endoplasmic reticulum resident protein 60, ERp60, grp60, HACBP) is encoded by the CALR (also known as CRTC) gene (Gene ID 811) in human. Calreticulin (CRT), BiP (GRP78 or heat shock 70 kDa protein 5), and protein disulphide isomerase (PDI) are ER lumen folding factors that, together with other PTM enzymes, assist the process of newly synthesized proteins before their release from ER. CRT, BiP, and PDI themselves are subject to posttranslational signal peptide removal after their synthesis, exposing E18, E19, and D18 at the newly formed N-terminus, respectively. The N-terminus D and E residues of the mature proteins are permissive to arginine-tRNA-protein transferase 1/ATE1-catalyzed arginylation, forming arginylated mature proteins (R-CRT, R-BiP, R-PDI) with altered cellular localization to allow their participation in non-ER functions. Although only the basal levels of R-CRT and R-PDI, but not R-BiP, are generally detectible in non-stimulated cells, upregulated N-terminal arginylation of all three proteins is observed upon cytosolic dsDNA exposure and proteasomal inhibition, indicating a shared role in innate immune responses to invading microbes. In addition, ER stress induction by thapsigargin treatment synergistically boosts proteasomal inhibition-induced upregulation of cellular levels of R-CRT, R-BiP, and R-PDI, suggesting that ER stress accelerates the supply of ER lumenal CRT, BiP, and DPI for N-terminal arginylation.

Specificity

This rabbit polyclonal antibody specifically detected the immunogen peptide, but not the control peptide without arginylation at the N-terminal Glu18 (Cha-Molstad, H., et al. (2015). Nat. Cell Biol. 17(7):917-929).

Immunogen

Epitope: N-terminus
Linear peptide corresponding to the N-terminal sequence of arginylated mature human Calreticulin.

Application

Detect arginylated mature calreticulin using this rabbit polyclonal Anti-Calreticulin, arginylated (Nt-Glu18), Cat. No. ABS1671, validated for use in ELISA, Immunocytochemistry, and Western Blotting.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Calreticulin Nt-Glu18 arginylation induction in (18-hr 3 µM MG132 and 200 nM thapsigargin) treated HeLa cells (Courtesy of Yong Tae Kwon, Ph.D. , Seoul National University, Korea).

Western Blotting Analysis: 0.2 µg/mL from a representative lot detected Calreticulin Nt-Glu18 arginylation induction in (18-hr 3 µM MG132 and 200 nM thapsigargin) treated HeLa cells (Courtesy of Yong Tae Kwon, Ph.D. , Seoul National University, Korea).

ELISA Analysis: A representative lot detected the immunogen peptide, but not the control peptide without arginylation at the N-terminal Glu18 (Cha-Molstad, H., et al. (2015). Nat. Cell Biol. 17(7):917-929).

Western Blotting Analysis: A representative lot detected Calreticulin Nt-Glu18 arginylation induction upon arginine-tRNA-protein transferase 1 (ATE1) 1A7A isoform overexpression or transfection of various dsDNAs, including poly(dA:dT), in HeLa cells. Combined proteasome inhibition and ER stress induction by an 18-hr 10 µM MG132 and 100 nM thapsigargin treatment synergized the two drugs′ efficacy toward cellular Calreticulin Nt-Glu18 arginylation induction (Cha-Molstad, H., et al. (2015). Nat. Cell Biol. 17(7):917-929).
Research Category
Signaling

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Calreticulin Nt-Glu18 arginylation induction in 7.5 µg of lysate from (17-hr 3 µM MG132 and 200 nM thapsigargin) treated HEK293 cells.

Target description

~55 kDa observed. 46.6 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service