Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABS1610

Sigma-Aldrich

Anti-ERLIN-2/SPFH2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Erlin-2By similarity, Endoplasmic reticulum lipid raft-associated protein 2ImportedBy similarity, Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2By similarity, SPFH domain-containing protein 2, ERLIN-2/SPFH2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ERLIN2(11160)

General description

Erlin-2 (UniProt B5DEH2; also known as Endoplasmic reticulum lipid raft-associated protein 2, SPFH domain-containing protein 2, SPFH domain family member 2, Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2) is encoded by the Erlin2 (also known as RGD1309010, Spfh2) gene (Gene ID 290823) in rat species. Erlin2 (Spfh2) and the highly homologous Erlin1 (Spfh1) are ER proteins that hetero-oligomerize into a large complex of ∼2 MDa. The erlin1/2 complex associates with inositol 1,4,5-trisphosphate (IP(3)) receptors on ER membrane and is responsible for recruiting RNF170 upon IP(3) receptor activation for subsequent ubiquitination-dependent receptor downregulation.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of rat ERLIN-2/SPFH2.

Application

Research Category
Signaling
Research Sub Category
Signaling Neuroscience
This Anti-ERLIN-2 Antibody is validated for use in Western Blotting, Immunocytochemistry and Immunoprecipitation for the detection of SPFH2.
Western Blotting Analysis: A representative lot detected ERLIN2/SPFH2 in HeLa cell lysates with or without deglycosylation by Endo H treatment (Pearce, M.M., et al. (2007). J Biol Chem. 282(28):20104-20115)
Western Blotting Analysis: A representative lot detected ERLIN2/SPFH2 in Rat-1 & HeLa cell lysates and various rat tissue homogenates, as well as IP3Rs activation-dependent increase of co-precipitated ERLIN2/SPFH2 in IP3Rs IP from rat gonadotroph pituitary alpha T3-1 cells and rat pancreatic tissue (Pearce, M.M., et al. (2007). J Biol Chem. 282(28):20104-20115).
Immunocytochemistry Analysis: A representative lot detected ERLIN2/SPFH2 ER localization in HeLa cells (Pearce, M.M., et al. (2007). J Biol Chem. 282(28):20104-20115).
Immunoprecipitation Analysis: A representative lot immunoprecipitated ERLIN2/SPFH2, as well as ERLIN2/SPFH2-associated IP3R1 and ERAD pathway proteins from rat pituitary gonadotroph alpha T3-1 cells (Pearce, M.M., et al. (2007). J Biol Chem. 282(28):20104-20115).

Quality

Evaluated by Western Blotting in rat liver tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ERLIN-2/SPFH2 in 10 µg of rat liver tissue lysate.

Target description

~43 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 155mM NaCl, 50mM Tris-HCl Ph 7.4 with 0.05% sodium azide and 10% glycerol.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heidrun Steinle et al.
Journal of cell science, 134(12) (2021-06-22)
The receptor interacting serine/threonine kinase 2 (RIPK2) is essential for signal transduction induced by the pattern recognition receptors NOD1 and NOD2 (referred to collectively as NOD1/2). Upon NOD1/2 activation, RIPK2 forms complexes in the cytoplasm of human cells. Here, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service