Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS1519

Sigma-Aldrich

Anti-phospho-SIN1 (Thr86)

from rabbit, purified by affinity chromatography

Synonym(s):

Target of rapamycin complex 2 subunit MAPKAP1, TORC2 subunit MAPKAP1, Mitogen-activated protein kinase 2-associated protein 1, Stress-activated map kinase-interacting protein 1, SAPK-interacting protein 1, mSIN1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$182.00
100 μL
$429.00

$182.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$182.00
100 μL
$429.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$182.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

inhibition assay: suitable (peptide)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

phosphorylation (pThr86)

Gene Information

human ... MAPKAP1(79109)

General description

Target of rapamycin complex 2 subunit MAPKAP1 (UniProt: Q9BPZ7; also known as TORC2 subunit MAPKAP1, Mitogen-activated protein kinase 2-associated protein 1, Stress-activated map kinase-interacting protein 1, SAPK-interacting protein 1, mSIN1) is encoded by the MAPKAP1 (also known as MIP1, SIN1) gene (Gene ID: 79109) in human. mSIN1 is a subunit of mTORC2 regulates cell growth and survival in response to hormonal signals. It is ubiquitously expressed, with highest levels in heart and skeletal muscle. Six isoforms of SIN1 have been described that are produced by alternative splicing. All isoforms with the exception of isoform 4 can be incorporated into the mTORC2 complex. mTORC2 is shown to be activated by growth factors, but in contrast to mTORC1, it is nutrient-insensitive. mTORC2 phosphorylates serine 473 in Akt1, which facilitates the phosphorylation of Threonine 308 by PDK1, thereby allowing full activation of Akt. Genetic knockout of SIN1 is reported to abolish serine 473 phosphorylation of Akt and disrupts the RICTOR-mTOR interaction. mTORC2 also regulates the phosphorylation of SGK1 at serine 422. Within mTORC2, MAPKAP1 is required for complex formation and mTORC2 kinase activity. MAPKAP1 inhibits MAP3K2 by preventing its dimerization and auto-phosphorylation.

Specificity

This rabbit polyclonal antibody detects human Stress-activated MAP kinase-interacting protein 1 phosphorylated on Threonine 86.

Immunogen

KLH-conjugated linear peptide corresponding to 10 amino acids surrounding phosphorylated threonine 86 residue from human Stress-activated MAP kinase-interacting protein 1. The immunogen sequence is conserved in all isoforms except isoform 4.

Application

Anti-phospho-SIN1, (Thr86), Cat. No. ABS1519, is a rabbit polyclonal antibody that detects Target of rapamycin complex 2 subunit MAPKAP1 (SIN1) and has been tested for use in Western Blotting and Peptide Inhibition Assay.
Peptide Inhibition Analysis: A 1:1,000 dilution from a representative lot was used with HEK293 cells treated with Insulin for peptide block analysis.
Research Category
Signaling

Quality

Evaluated by Western Blotting in HEK293 treated with Insulin.

Western Blotting Analysis: A 1:500 dilution of this antibody detected phospho-SIN1, (Thr86) in lysates of HEK293 cells treated with Insulin.

Target description

~65 kDa observed; 59.12 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service