Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS13

Sigma-Aldrich

Anti-TRABID Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Ubiquitin thioesterase ZRANB1, TRAF-binding domain-containing protein, hTrabid, Zinc finger Ran-binding domain-containing protein 1, ZRANB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Gene Information

human ... ZRANB1(54764)

General description

TRAF-binding domain-containing protein (TRABID) is also known as Ubiquitin thioesterase ZRANB1 and Zinc finger Ran-binding domain-containing protein 1. TRABID specifically hydrolyzes ′Lys-29′-linked and ′Lys-33′-linked diubiquitin, as well as′Lys-63′-linked chains, but with less efficiency. TRABID positively regulates the Wnt signaling pathway, is involved in regulation of cell morphology, cytoskeletal organization, stress fiber dynamics and cell migration, and may also modulate TNF-alpha signaling.

Immunogen

Recombinant protein corresponding to the N-terminus of human TRABID.

Application

Detect TRAF-binding domain-containing protein using this rabbit polyclonal antibody, Anti-TRABID Antibody validated for use in western blotting & IP.
Immunoprecipitation Analysis: 59 µL from a representative lot immunoprecipitated TRABID from 500 µg of HeLa cell lysate. Immunopreciptated protein was then probed with a 1:170 dilution of the same antibody in WB.
Research Category
Metabolism
Research Sub Category
General Post-translation Modification

Quality

Evaluated by Western Blotting in untransfected and TRABID transfected HEK293 cell lysate.

Western Blotting Anlaysis: A 1:1,000 dilution from a representative lot detected TRABID in TRABID transfected HEK293 cell lysate.

Target description

~80 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service