Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS1073

Sigma-Aldrich

Anti-Dio3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Type III iodothyronine deiodinase, 5DIII, DIOIII, Type 3 DI, Type-III 5′-deiodinase, Dio3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$472.00

$472.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$472.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$472.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

primate (based on 100% sequence homology)

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable

GenBank accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DIO3(1735)

General description

The enzyme DIO3 is a critical enzyme in thyroid hormone metabolism. It converts the thyroid hormone T4 into the inactive form RT3 and it also converts the thyroid hormone T3 into the inactive form T2. It is thought that DIO3 plays a role in preventing premature exposure of the developing fetus to adult levels of thyroid hormones. DIO3 also regulates the levels of fetal thyroid hormones during gestation, thus DIO3 is critical for proper development and metabolism of the fetus during embryological development. DIO3 is expressed in the placenta and in fetal tissues, and DIO3 is localized to the cell membrane and endosomal membranes. DIO3 is also often expressed in neoplastic tissues and in particular basal cell carcinomas.

Immunogen

BSA-conjugated linear peptide corresponding to the extracellular domain of human Dio3.
Epitope: Extracellular domain

Application

Anti-Dio3 Antibody is an antibody against Dio3 Antibody for use in Immunohistochemistry, Immunofluorescence.
Immunohistochemistry Analysis: A 1:100 dilution from a representative lot detected Dio3 in human placenta.
Immunohistochemistry Analysis: A representative lot detected Dio3 in Hepatic tissue (Huang, S.A., et al. (2001). Journal of Clinical Endocrinology & Metabolism. 87(10):4457–4461).
Immunohistochemistry Analysis: A representative lot detected Dio3 in human uterus, placenta, umbilical cord, fetal epithelial, and endometrium (Huang, S.A., et al. (2003). Journal of Clinical Endocrinology & Metabolism. 88(3):1384-1388).
Research Category
Signaling
Research Sub Category
Signaling Neuroscience

Quality

Evaluated by Immunohistochemistry in human uterus tissue.

Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected Dio3 in human uterus tissue.

Target description

34 kDa calculated

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service