Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS1025

Sigma-Aldrich

Anti-NUB1 Antibody

serum, from rabbit

Synonym(s):

NEDD8 ultimate buster 1, Negative regulator of ubiquitin-like proteins 1, Renal carcinoma antigen NY-REN-18

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

GenBank accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NUB1(51667)

General description

NUB1 is a critical control protein for the ubiquitin proteasome system in that it regulates NEDD8 activity by specifically targeting NEDD8, ubiquitin D (FAT10), and their conjugates to the proteasome for degradation. There are two isoforms, isoform one, (NUB1L) is more efficient at NEDD8 recruitment than isoform 2. Interestingly NUB1 is localized primarily in the nucleus of resting cells and its expression is wide spread from leukocytes in the blood to liver cells and even skeletal muscle cells. Also interesting is that NUB1 was identified early as an auto-antigen in renal carcinomas. NUB1 appears to operate via binding to Asn-51 on NEDD8 which complexes with the UFD1-NPL4 complex that facilitates degradation via the proteasome.

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminus of human NUB1.

Application

Anti-NUB1 Antibody | ABS1025 is an antibody against NUB1 Antibody for use in Western Blotting, Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected NUB1 in human retina tissue.
Western Blotting Analysis: A representative lot detected NUB1 in NUB1-transfected HEK293 lysate (Schmidtke, G., et al. (2006). J. Biol. Chem. 281:20045-20054).
Western Blotting Analysis: A representative lot detected NUB1 in retinal extracts, Y79, SH-SY5Y, and Hela lysate (Van der Spuy, J., et al. (2003). IOVS. 44(12):5396-5403).
Western Blotting Analysis: A representative lot detected NUB1 in SK-N-SH cell transfected with NUB1-FLAG lysate (Bett, J.S., et al. (2012). PLoS ONE. 7(2):e30866).
Immunohistochemistry Analysis: A representative lot detected NUB1 in retina, cornea, sclera, choroid, iris, and cillary body of developing human eye tissue (Van der Spuy, J., et al. (2003). IOVS. 44(12):5396-5403).

Quality

Evaluated by Western Blotting in SH-SY5Y cell lysate.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected NUB1 in 10 µg of SH-SY5Y cell lysate.

Target description

~70 kDa observed. Uncharacterized band(s) may appear in some lysates.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service