ABN994
Anti-ZNT3 pAb Antibody
from rabbit, purified by affinity chromatography
Synonym(s):
Zinc transporter 3, Solute carrier family 30 member 3, ZnT-3
About This Item
Recommended Products
biological source
rabbit
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
monkey, human, mouse
technique(s)
electron microscopy: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... SLC30A3(7781)
General description
Specificity
Immunogen
Application
Neuroscience
Ion Channels & Transporters
Western Blotting Analysis: A representative lot detected ZnT-3 expression in brain lysates from wild-type, but not Znt3-knockout mice, while reduced ZnT-3 level was seen in brain tissues from Znt3+/- mice (Cole, T.B., et al. (1999). Proc. Natl. Acad. Sci. U. S. A. 96(4):1716-17121).
Western Blotting Analysis: A representative lot detected exogenously expressed murine ZnT-3 in transfected BHK baby hamster kidney fibroblasts and endogenous ZnT-3 in mouse brain, but not testis or kidney, tissue homogenates. No cross-reactivity was seen toward murine ZnT-1 or ZnT-2 (Palmiter, R.D., et al. (1996). Proc. Natl. Acad. Sci. U. S. A. 93(25):14934-14939).
Immunohistochemistry Analysis: A representative lot detected ZnT-3 immunoreactivity in the hippocampus inner molecular layer (IML) of 4% paraformaldehyde-/0.025% glutaraldehyde-fixed brain sections from mice undergoing spontaneous recurrent seizures (SRS) induction by pilocarpine treatment (Chi, Z.H., et al. (2008). Neurochem. Int. 52(7):1305-1309).
Immunohistochemistry Analysis: A representative lot detected ZnT-3 immunoreactivity in brain regions of wild-type, but not Znt3-knockout mice, while reduced ZnT-3 immunoreactivity was seen in brain tissues from Znt3+/- mice using 4% paraformaldehyde-fixed frozen brain sections (Cole, T.B., et al. (1999). Proc. Natl. Acad. Sci. U. S. A. 96(4):1716-17121).
Immunohistochemistry Analysis: A representative lot detected ZnT-3 immunoreactivity in the zinc-containing mossy fiber (MF) projections in mouse and monkey hippocampus using 4% paraformaldehyde-fixed frozen brain sections (Wenzel, H.J., et al. (1997). Proc. Natl. Acad. Sci. U. S. A. 94(23):12676-12681).
Immunohistochemistry Analysis: A representative lot immunostained Zn-rich mossy fiber projections emanating from dentate gyrus granule cell neurons as well as projections from hippocampal and cortical neurons in 4% paraformaldehyde-fixed frozen mouse brain sections (Palmiter, R.D., et al. (1996). Proc. Natl. Acad. Sci. U. S. A. 93(25):14934-14939).
Immunofluorescence Analysis: A representative lot immunostained mossy fiber (MF) sprouting in the hippocampus inner molecular layer (IML) by fluorescent immunohistochemistry staining of 4% paraformaldehyde-fixed free-floating brain sections from pilocarpine-induced epileptic mice with or without medial ganglionic eminence (MGE) progenitor cell grafts (Hunt, R.F., et al. (2013). Nat. Neurosci. 16(6):692-697).
Immunofluorescence Analysis: A representative lot detected ZnT-3 immunoreactivity in Aβ-positive senile plaques (SP) by dual fluorescent immunohistochemistry staining of 4% paraformaldehyde-fixed, paraffin-embedded Alzheimer′s diseased human cerebral cortex sections (Zhang, L.H., et al. (2008). Brain Res. Bull. 77(1):55-60).
Electron Microscopy: A representative lot immunostained mossy fiber bouton (MFB) in the hippocampus inner molecular layer (IML) of 4% paraformaldehyde-/0.025% glutaraldehyde-fixed frozen brain sections from mice undergoing spontaneous recurrent seizures (SRS) induction by pilocarpine treatment (Chi, Z.H., et al. (2008). Neurochem. Int. 52(7):1305-1309).
Electron Microscopy: A representative lot detected ZnT-3 immunoreactivity at the terminals of glutamatergic projections in mouse and monkey hippocampus by EM using 4% paraformaldehyde-fixed frozen brain sections (Wenzel, H.J., et al. (1997). Proc. Natl. Acad. Sci. U. S. A. 94(23):12676-12681).
Quality
Western Blotting Analysis: A 1:500 dilution of this antibody detected ZNT3 in 10 µg of Alzheimer′s diseased human brain tissue lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service