Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABN643

Sigma-Aldrich

Anti-LRRTM2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Leucine-rich repeat transmembrane neuronal protein 2, Leucine-rich repeat neuronal 2 protein, LRRTM2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$354.00

$354.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$354.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$354.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Leucine-rich repeat transmembrane neuronal protein 2, also known as Leucine-rich repeat neuronal 2 protein, and encoded by the gene name LRRTM2 or LRRN2, is involved in the development and maintenance of excitatory synapse in the vertebrate nervous system. The Leucine-rich repeat transmembrane neuronal proteins (LRRTMs) are differentially expressed in the nervous system and were recently found to instruct presynaptic and mediate postsynaptic glutamatergic differentiation, with LRRTM1 and LRRTM2 most potent at inducing presynaptic differentiation. Each LRRTM protein is a type I transmembrane containing ten extracellular leucine-rich repeats and a short intracellular tail and has a developmentally regulated pattern distinct from all others. LRRTM2 interacts with PSD-95 and regulates the surface expression of AMPA receptors. LRRTM2 also functions as a neurexin ligand, binding both Neurexin 1-alpha and -beta, suggesting that LRRTM2-Neurexin1 interaction plays a critical role in regulatory excitatory synapse development

Immunogen

Epitope: cytoplasmic domain
KLH-conjugated synthetic peptide corresponding to the cytoplasmic domain of human LRRTM2.

Application

Anti-LRRTM2 Antibody detects level of LRRTM2 & has been published & validated for use in LRRTM2.
Immunohistochemistry Analysis: 2.5 μg/mL from a representative lot detected LRRTM2 in human brain tissue.





Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blotting in SK-N-SH cell lysate with and without blocking peptide.

Western Blotting Analysis: 1 μg/mL of this antibody detected LRRTM2 in SK-N-SH cell lysate.

Target description

~65 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service