Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABN446

Sigma-Aldrich

Anti-Capicua Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Protein capicua homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

ox (based on 100% sequence homology), hamster (based on 100% sequence homology), mouse (based on 100% sequence homology), chimpanzee (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CIC(23152)

General description

Protein capicua homolog (CIC) , also known as Capicua Transcriptional Repressor, Capicua Homolog, and encoded by the gene CIC, is a transcriptional repressor which plays a role in development of the central nervous system (CNS). CIC is a new subfamily of the HMG-box superfamily and the gene′s expression is predominantly restricted to immature granule cells in the cerebellum, hippocampus and olfactory bulb in the CNS, and thus the gene is implicated in CNS development, in particular in granule cell development. CIC appears to be central to cerebellar development and ErbB signaling as well. Recent research has demonstrated that CIC is highly expressed in certain cancers including medulloblastomas, but also the loss of expression may serve as a biomarker for other cancer types such as oligodendrogliomas.

Immunogen

Epitope: Near N-terminal
KLH-conjugated linear peptide corresponding to human Capicua near the N-terminal.

Application

Immunohistochemistry Analysis: A 1:50-1,000 dilution from a representative lot detected Capicua in human cerebral cortex tissue.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-Capicua Antibody is validated for use in Western Blotting for the detection of Capicua.

Quality

Evaluated by Western Blotting in human cerebellum tissue lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Capicua in 10 µg of human cerebellum tissue lysate.

Target description

~160 kDa observed. Long isoform can be detected at ~250kDa with longer exposure, in some lysates (Kim, E., et al. (2013). Genea Dev. 27:590-595).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shih-Chiang Huang et al.
The American journal of surgical pathology, 40(5), 645-655 (2016-01-07)
Angiosarcoma (AS) is a rare sarcoma subtype showing considerable clinicopathologic and genetic heterogeneity. Most radiation-induced AS show MYC gene amplifications, with a subset of cases harboring KDR, PTPRB, and PLCG1 mutations. Despite recent advances, the genetic abnormalities of most primary
Severa Bunda et al.
Nature communications, 10(1), 661-661 (2019-02-10)
Capicua (CIC) is a transcriptional repressor that counteracts activation of genes downstream of receptor tyrosine kinase (RTK)/Ras/ERK signaling. It is well-established that tumorigenesis, especially in glioblastoma (GBM), is attributed to hyperactive RTK/Ras/ERK signaling. While CIC is mutated in other tumors
Severa Bunda et al.
Molecular cancer research : MCR, 18(5), 774-786 (2020-02-08)
Capicua (CIC) is a transcriptional repressor that counteracts activation of genes in response to receptor tyrosine kinase (RTK)/Ras/ERK signaling. Following activation of RTK, ERK enters the nucleus and serine-phosphorylates CIC, releasing it from its targets to permit gene expression. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service