Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABN425

Sigma-Aldrich

Anti-DISC1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Disrupted in schizophrenia 1 homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Disc1(244667)

General description

Disrupted in schizophrenia 1 protein (DISC1) is involved in the regulation of multiple aspects of embryonic and adult neurogenesis. DISC1 may play a role as a modulator of the AKT-mTOR signaling pathway controlling the tempo of the process of newborn neurons integration during adult neurogenesis, including neuron positioning, dendritic development and synapse formation. DISC1 is shown to inhibit the activation of AKT-mTOR signaling upon interaction with CCDC88A. DISC1 is required for neural progenitor proliferation in the ventrical/subventrical zone during embryonic brain development and in the adult dentate gyrus of the hippocampus. Additionally, DISC1 is suggested to regulate the migration of early-born granule cell precursors toward the dentate gyrus during the hippocampal development. DISC1 has been shown to participates in the Wnt-mediated neural progenitor proliferation as a positive regulator and may have a role, together with PCNT, in the microtubule network formation. DICS1 is highly expressed in the dentate gyrus of the hippocampus. It is also expressed in the temporal and parahippocampal cortices and cells of the white matter. Expression rises within the dentate gyrus and temporal cortex from the neonatal period to infancy, declines markedly in adolescence, and declines further with aging.

Immunogen

Recombinant protein corresponding to mouse DISC1.

Application

Anti-DISC1 Antibody detects level of DISC1 & has been published & validated for use in DISC1.
Immunohistochemistry Analysis: A representative lot detected DISC1 in mouse hippocampus tissue (Kuroda, K., et al. (2011). Human Mole. Gene. 20(23):4666-4683).
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in mouse wild type P0 brain tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected DISC1 in 10 µg of mouse wild type P0 brain and mouse knock out P0 brain tissue lysate.

Target description

~ 94 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cana Park et al.
Molecular brain, 9(1), 69-69 (2016-07-03)
In neuronal axons, the ratio of motile-to-stationary mitochondria is tightly regulated by neuronal activation, thereby meeting the need for local calcium buffering and maintaining the ATP supply. However, the molecular players and detailed regulatory mechanisms behind neuronal mitochondrial movement are
Yeongjun Suh et al.
Molecular neurobiology, 56(10), 6725-6735 (2019-03-28)
Disrupted-in-Schizophrenia 1 (DISC1) is a scaffold protein implicated in various psychiatric diseases. Dysregulation of the dopamine system has been associated with DISC1 deficiency, while the molecular mechanism is unclear. In this study, we propose a novel molecular mechanism underlying the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service