Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABN353

Sigma-Aldrich

Anti-SAPAP2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

SAP90, PSD-95-associated protein 2, Disks large-associated protein 2, DLGAP2, DAP-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$354.00

$354.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$354.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$354.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DLGAP2(9228)

General description

SAP90/PSD-95-associated protein 2 (SAPAP2, also known as DLGAP2) is a member of a protein family whose members specifically interact with PSD-95/SAP90, a membrane-associated guanylate kinase localized at postsynaptic density (PSD) in neuronal cells. Like the other SAPAP proteins, SAPAP2 is thought to be an adaptor protein that also interacts with different synaptic scaffolding proteins, cytoskeletal and signaling components, such as focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2).

Specificity

At least three isoforms of SAPAP2 are known to exist. This SAPAP2 antibody will not cross-react with other SAPAP proteins.

Immunogen

KLH-conjugated linear peptide corresponding to human SAPAP2.

Application

Immunohistochemistry Analysis: 5 µg/mL of this antibody from a representative lot detected SAPAP2 in human brain tissue.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
This SAPAP2 antibody is validated for use in WB & IHC for the detection of the SAPAP2 protein.

Quality

Evaluated by Western Blot in L1210 cell lysate.

Western Bloting Analysis: 1 µg/mL of this antibody detected SAPAP2 in 15 µg of L1210 cell lysate.

Target description

~114 kDa observed. Uncharacterized bands may be observed.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS and 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
L1210 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service