Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABN345

Sigma-Aldrich

Anti-Slitrk2 Antibody

from rabbit

Synonym(s):

SLIT and NTRK-like protein 2, CXorf2, SLITL1, KIAA1854, slit-like 1, SLIT and NTRK-like protein 2, slit and trk like gene 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLITRK2(84631)

General description

SLIT and NTRK-like family 2 (Slitrk2) is a member a protein family consisting of six homologous transmembrane proteins (Slitrk1-6) that share two conserved leucine-rich repeat domains in the extracellular domain and have significant homology to Slit, a secreted axonal growth-controlling protein. These proteins are also homologous to trk neurotrophin receptors in their intracellular domains. Expression of Slitrk proteins is highly restricted to neural and brain tumor tissues, but varies within the family. For example, Slitrk2 is expressed primarily in neurons in the ventricular layer of the brain. Like every other Slitrk protein except Slitrk1, overexpression of Slitrk2 inhibited neurite outgrowth in cultured neurons. At least two isoforms of this protein are known to exist.

Immunogen

KLH-conjugated linear peptide corresponding to human Slitrk2.

Application

Immunohistochemistry Analysis: 2.5 µg/mL from a representative lot detected Slitrk2 in human brain tissue.
Research Category
Neuroscience

Neuroscience
Research Sub Category
Developmental Neuroscience

Neurofilament & Neuron Metabolism
This Slitrk2 antibody is validated for use in WB & IHC for the detection of the Slitrk2 protein.

Quality

Evaluated by Western Blot in rat brain tissue lysate.

Western Blot Analysis: 2 µg/mL of this antibody detected Slitrk2 in 15 µg of rat brain tissue lysate.

Target description

~98 kDa observed

Physical form

Antigen Affinity Purified
Format: Purified
Purified rabbit polyclonal in buffer containing PBS with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain tissue lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service