SAP90/PSD-95-associated protein 4 (SAPAP4, also known as DLGAP4) is a member of a protein family whose members specifically interact with PSD-95/SAP90, a membrane-associated guanylate kinase localized at postsynaptic density (PSD) in neuronal cells. Like the other SAPAP proteins, SAPAP4 is thought to be an adaptor protein that also interacts with different synaptic scaffolding proteins, cytoskeletal and signaling components, such as focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2). SAPAP4 mRNA is targeted to cell bodies in a similar manner to SAPAP1 and -2, whereas SAPAP3 mRNA is detected mainly in cell bodies. At least three isoforms of SAPAP4 are known to exist.
Specificity
This SAPAP4 antibody will not cross-react with other SAPAP proteins.
Immunogen
KLH-conjugated linear peptide corresponding to human SAPAP4.
Application
Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected SAPAP4 in rat hippocampus tissue.
This SAPAP4 antibody is validated for use in WB & IHC for the detection of the SAPAP4 protein.
Quality
Evaluated by Western Blot in SK-N-SH cell lysate.
Western Blot Analysis: 1 µg/mL of this antibody detected SAPAP4 in 15 µg/mL of SK-N-SH cell lysate.
Target description
~138 kDa observed. The calculated molecular weight of this protein is 109 kDa, but can be observed at ~138 kDa due to post translational modification.
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.