Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABN2258

Sigma-Aldrich

Anti-poly-(LPAC)

serum, from rabbit

Synonym(s):

(Leu-Pro-Ala-Cys)Exp

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$179.00
100 μL
$452.00

$179.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$179.00
100 μL
$452.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$179.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

target post-translational modification

unmodified

General description

RAN, a small GTP binding protein, belonging to the RAS superfamily is involved in the translocation of RNA and proteins through the nuclear pore complex and participate in the control of DNA synthesis and cell cycle progression. LPAC (Leu-Pro-Ala-Cys) RAN proteins that are primarily localized to cytoplasmic region are reported to accumulate in the gray matter region of the brain of subjects with myotonic dystrophy type 2 (DM2). LPAC cytoplasmic aggregates are found in neurons, astrocyte, and glia. LPAC proteins are shown to be toxic to cells independent of RNA effects and are found in regions of the brain that display pathologic changes. Studies have shown that tetranucleotide DM2 CCTG expansion mutation is bidirectionally transcribed and the resulting RNAs are RAN translated and produce tetrapeptide expansion proteins with LPAC from the sense strand and that these proteins accumulate in DM2 patient brains. (Ref.: Zu, T., et al. (2017). Neuron 95 (8); 1292-1305).

Specificity

This rabbit polyclonal antibody specifically detects proteins with poly-(LPAC).

Immunogen

A synthetic peptide corresponding to the (LPAC)EXP repeat motif.
Epitope: unknown

Application

Anti-poly-(LPAC), Cat. No. ABN2258, is a highly specific rabbit polyclonal antibody that targets poly-(LPAC) and has been tested for use in Immunohistochemistry, Immunofluorescence, and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis (WB): 1:1000 of this antibody detected poly-(LPAC) HEK293T-poly-(LPAC) cells, but not in control cells..

Western Blotting Analysis: A representative lot detected poly-(LPAC) in Western Blotting applications (Banez-Coronel, M., et. al. (2015). Neuron. 88(4):667-77).

Immunofluorescence Analysis: A representative lot detected poly-(LPAC) in Immunofluorescence applications (Banez-Coronel, M., et. al. (2015). Neuron. 88(4):667-77).

Immunohistochemistry Analysis: A representative lot detected poly-(LPAC) in Immunohistochemistry applications (Banez-Coronel, M., et. al. (2015). Neuron. 88(4):667-77).

Quality

Evaluated by Western Blotting in HEK293T cells transfected with poly-(LPAC).


Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected poly-(LPAC) in HEK293T cells transfected with poly-(LPAC), but not in control cells.

Target description

~25 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Rabbit polyclonal antiserum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah Rösing et al.
Nature communications, 15(1), 1534-1534 (2024-02-21)
Myotonic dystrophy type 2 (DM2) is a tetranucleotide CCTG repeat expansion disease associated with an increased prevalence of autoimmunity. Here, we identified an elevated type I interferon (IFN) signature in peripheral blood mononuclear cells and primary fibroblasts of DM2 patients

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service