Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABN1718

Sigma-Aldrich

Anti-SLC4A11 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Sodium bicarbonate transporter-like protein 11, Bicarbonate transporter-related protein 1, NaBC1, Sodium borate cotransporter 1, Solute carrier family 4 member 11

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$429.00

$429.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$429.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$429.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, bovine, rabbit, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

Sodium bicarbonate transporter-like protein 11 (UniProt A2AJN7; also known as Bicarbonate transporter-related protein 1, NaBC1, Sodium borate cotransporter 1, Solute carrier family 4 member 11) is encoded by the Slc4a11 (also known as BTR1) gene (Gene ID 269356) in murine species. Bicarbonate (HCO3-) transporters are the principal regulators of pH in cells and play a vital role in acid-base movement in various tissues. There exist four types of HCO3- transporters, namely Cl-/-HCO3- exchangers, Na+/HCO3- cotransporters (NBCs), a K+/HCO3- cotransporter, and a Na+-driven Cl-/-HCO3- exchanger. SLC4A11 is a ubiquitously expressed 14-transmembrane protein existing as dimers within the plasma membrane. Originally classified as bicarbonate transporter, later stiudy conducted with cultured primary bovine corneal endothelial cells (BCECs) indicated that SLC4A11 mediates Na+-coupled hydroxide (OH-) transport, but not borate (B(OH)4-) or bicarbonate (HCO3-) transport in corneal endothelium. SLC4A11 gene mutations in human are linked to endothelial dystrophies, including CDPD (Corneal dystrophy and perceptive deafness), CHED2 (Corneal dystrophy, endothelial 2, autosomal recessive), and FECD4 (Corneal dystrophy, Fuchs endothelial, 4).

Specificity

Specificity of this polyclonal antibody was verified by antigen peptide blocking of target band detection in Western blotting application (Jalimarada, S.S., et al. (2013). Invest. Ophthalmol. Vis. Sci. 54(6):4330-4340). Epitope sequence is conserved between murine and human (UniProt A2AJN7 and Q8NBS3) and present in all four human SLC4A11 spliced isoforms reported by UniProt.

Immunogen

Linear peptide corresponding to a sequence near the C-terminus of mouse SLC4A11.

Application

Immunocytochemistry Analysis: A 1:80 dilution from a representative lot detected the expression of exogenously transfected human SLC4A11 in PS120 Chinese hamster lung fibroblasts (Courtesy of Professor Joseph A. Bonanno, O.D., PhD, Indiana University, Bloomington, IN).
Western Blotting Analysis: A representative lot detected SLC4A11 in cultured human and bovine corneal endothelial cell lysates, as well as in isolated rabbit and bovine corneal endothelium extracts (Jalimarada, S.S., et al. (2013). Invest. Ophthalmol. Vis. Sci. 54(6):4330-4340).
Western Blotting Analysis: A representative lot detected siRNA-mediated SLC4A11 downregulation in cultured bovine corneal endothelial cells (Jalimarada, S.S., et al. (2013). Invest. Ophthalmol. Vis. Sci. 54(6):4330-4340).
Immunocytochemistry Analysis: A representative lot detected predominant lateral membrane localization of SLC4A11 in isolated rabbit corneal endothelium, while peripheral staining was evident in cultured bovine corneal endothelial cells, by fluorescent immunocytochemistry staining of 2% paraformaldehyde-fixed, 0.2 % saponin-permeabilized cells (Jalimarada, S.S., et al. (2013). Invest. Ophthalmol. Vis. Sci. 54(6):4330-4340).
This Anti-SLC4A11 Antibody is validated for use in Western Blotting, Immunocytochemistry for the detection of SLC4A11.

Quality

Evaluated by Western Blotting in mouse eye tissue lysate.

Western Blotting Analysis: 5.0 µg/mL of this antibody detected SLC4A11 in 10 µg of mouse eye tissue lysate.

Target description

~110 kDa observed. Target band size appears larger than the calculated molecular weights of 96.75 kDa (mouse) and 99.58/45.76/98.18/103.1 kDa (human isoform 1/2/3/4) due to glycosylation. Uncharacterized band(s) may appear in some lysates.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service