ABN1387
Anti-Beta-glucocerebrosidase (GBA)
serum, from mouse
Synonym(s):
Glucosylceramidase, Acid beta-glucosidase, Alglucerase, Beta-GC, Beta-glucocerebrosidase, D-glucosyl-N-acylsphingosine glucohydrolase, Imiglucerase
Select a Size
$414.00
Select a Size
About This Item
$414.00
Recommended Products
biological source
mouse
Quality Level
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
technique(s)
ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... GBA(2629)
General description
Specificity
Immunogen
Application
Western Blotting Analysis: A representative lot detected His-tagged recombinant full-length human mature beta-glucocerebrosidase (GBA) (Courtesy of Dr. Cláudia Sossai Soares and Paulo Lee Ho).
ELISA Analysis: A representative lot detected purified recombinant human beta-glucocerebrosidase (GBA) (Novo, J.B., et al. (2010). Mol. Biotechnol. 46(3):279-286).
Western Blotting Analysis: A representative lot detected bacterially expressed unglycosylated recombinant human beta-glucocerebrosidase (GBA), glycosylated human GBA expressed and secreted by COS-7 transfectants, as well as endogenous GBA in HeLa and ECV304 cell lysates (Novo, J.B., et al. (2010). Mol. Biotechnol. 46(3):279-286).
Quality
Western Blotting Analysis: A 1,000 dilution of this antiserum detected beta-glucocerebrosidase (GBA) in 10 µg of SK-N-SH human neuroblastoma cell lysate.
Target description
Physical form
Other Notes
Not finding the right product?
Try our Product Selector Tool.
recommended
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service