Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABN1029

Sigma-Aldrich

Anti-CHD-8 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Chromodomain-helicase-DNA-binding protein 8, CHD-8, ATP-dependent helicase CHD8, Axis duplication inhibitor, Duplin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CHD8(57680)

General description

CHD-8 is a DNA helicase that acts as a chromatin remodeling factor as well as a transcriptional regulator. CHD-8 acts as a transcription repressor by remodeling chromatin structure and recruiting histone H1 to various genes such as p53. CHD-8 regulates beta catenin activity by suppressing the promoter activity of several beta catenin targeted genes. CHD-8 also suppresses STAT3 transcriptional activation in a similar fashion. CHD-8 is a component of multiple chromatin complexes and localizes to the nucleus and active chromatin. Interestingly mutations in CHD-8 are associated with a form of autism, Autism 18 as well as some forms of developmental delay and cognitive impairment, demonstrating CHD-8’s potential importance in developmental brain function, and behavior.

Immunogen

Recombinant protein corresponding to rat CHD-8.

Application

Anti-CHD-8 | ABN1029 is an antibody against CHD-8 for use in Western Blotting, Immunocytochemistry, Immunoprecipitation.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling
Western Blotting Analysis: A representative lot detected CHD-8 in A431 cell lysate (Sakamoto, I., et al. (2000). JBC. 275:32871-32878).
Immunoprecipitation Analysis: A representative lot immunoprecipitated CHD-8 in β-catenin cell lysate (Sakamoto, I., et al. (2000). JBC. 275:32871-32878).
Immunocytochemistry Analysis: A representative lot detected CHD-8 in Lcells and SW480 cells (Sakamoto, I., et al. (2000). JBC. 275:32871-32878).

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected CHD-8 in 10 µg of HepG2 cell lysate.

Target description

~290 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service