Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABN1008

Sigma-Aldrich

Anti-ENTPD1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Ectonucleoside triphosphate diphosphohydrolase 1, NTPDase 1, Ecto-ATP diphosphohydrolase 1, Ecto-ATPDase 1, Ecto-ATPase 1, Ecto-apyrase, Lymphoid cell activation antigen, CD39, ENTPD1, NTPDase1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

primate (based on 100% sequence homology)

technique(s)

flow cytometry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ENTPD1(953)

General description

Ectonucleoside triphosphate diphosphohydrolase 1 (EC:3.6.1.5; UniProt P49961; also known as CD39, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPDase 1, Ecto-ATPase 1, Lymphoid cell activation antigen, NTPDase 1) is encoded by the ENTPD1 (also known as CD39, SPG64) gene (Gene ID 953) in human. CD39 is a cell surface nucleotide-metabolizing enzyme that hydrolyzes extracellular purine NTP and NDP. CD39 is a putative marker for B-cell activation and its expression is also observed on NK-cells, dendritic cells, T-cells, and vascular endothelial cells. CD39 is a known inhibitor of ADP-induced platelet aggregation and may also play a role in angiogenesis. E. coli infection is reported to induce CD39 promoter activity in macrophages, resulting in an upregulated CD39 expression during sepsis in mice, while CD39 knockdown increases mortality among mice with polymicrobial sepsis.

Specificity

Expeccted to react with all six spliced isoforms, including the Vascular and the Placenta I&II isoforms.

Immunogen

Epitope: extracellular domain
KLH-conjugated linear peptide corresponding to the extracellular domain of human ENTPD1.

Application

Research Category
Neuroscience
Research Sub Category
Developmental Signaling
This Anti-ENTPD1 Antibody is validated for use in Western Blotting, Flow Cytometry for the detection of ENTPD1.
Western Blotting Analysis: 2.0 µg/mL from a representative lot detected ENTPD1 in 10 µg of human spleen and Jurkat cell lystaes.
Flow Cytometry Analysis: 1.0 µg of this antibody from a representative lot detected ENTPD1 in 1X10E6 Jurkat cells.

Quality

Evaluated by Western Blotting in human placenta cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected ENTPD1 in 10 µg of human placenta cell lysate.

Target description

~65 kDa observed when using human placenta tissue lysates. Glycosylated target band(s) will appear larger than the calculated molecular weights of 58.0 kDa (Isoform 1/Vascular), 58.7 kDa ((Isoform 2/Placenta I), 34.2 kDa (Isoform 3/Placenta II), 42.7 kDa (Isoform 4), 46.2 kDa (Isoform 5), and 59.3 kDa (Isoform 6). Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service