Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABE989

Sigma-Aldrich

Anti-Ptx1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Pituitary homeobox 1, Hindlimb-expressed homeobox protein backfoot, Homeobox protein P-OTX, Homeobox protein PITX1, Paired-like homeodomain transcription factor 1, Pituitary OTX-related factor, Ptx1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

technique(s)

ChIP: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Pitx1(18740)
rat ... Pitx1(113983)

General description

Pituitary homeobox 1 (Ptx1; also known as Paired-like homeodomain transcription factor 1, Pituitary homeobox 1, or Hindlimb-expressed homeobox protein backfoot) is a nuclear transcription factor encoded by the Pitx1 (also known as Bft, Potx, Ptx1) gene in mouse. Ptx1 is reported to stimulate LHbeta gene promoter activity either alone or synergistically with the orphan nuclear receptor steroidogenic factor-1 (SF-1) and the early growth response gene 1 (Egr-1). Ptx1 is expressed in the hindlimb, but not the forelimb, of developing mouse embryos, and reduced hindlimb size and loss of hindlimb-specific features are seen in Pitx1-knockout mice.

Immunogen

Epitope: Near N-terminus
MBP-tagged recombinant protein corresponding to mouse Ptx1 near the N-terminus.

Application

Anti-Ptx1 Antibody is an antibody against Ptx1 for use in Western Blotting, Immunohistochemistry, Immunocytochemistry and Chromatin Immunoprecipitation (ChIP).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
Western Blotting Analysis: A 1:250 dilution from a representative lot detected Ptx1 in 10 µg of GH3 cell lysate.
Immunohistochemistry Analysis: A representative lot detected Ptx1 in adult and developing pituitary (Lanctot, C., et al. (1999) Endocrinology. 140(3):1416-1422).
Chromatin Immunoprecipitation Analysis: A representative lot immunoprecipitated Ptx1 in AtT-20 cells (Langlais, D., et al. (2011). Molecular Endocrinology. 25(2): 348–359).

Quality

Evaluated by Western Blotting in αT3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Ptx1 in 100 µg of αT3 cell lysate.

Target description

~48 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Affinity
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service