Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABE418

Sigma-Aldrich

Anti-phospho Histone H2A Antibody (Thr120)

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H2A type 1, H2A.1, Histone H2A/p

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

technique(s)

dot blot: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pThr120)

Gene Information

General description

Histone H2A is one of four components of the core nucleosomal structure. The nucleosome represents a unit of chromatin in which DNA is wrapped around a histone octamer. Histones undergo a number of post-translational modifications (PTM) in response to various stimuli that may induce changes in the structure of the nucleosome and hide or expose DNA sequences. Histone H2A, in particular, undergoes acetylation on lysine 5 by the Tip60 enzyme which may promote unfolding of chromatin and transcription. In addition, H2A is phosphorylated on serine 1 and serine 139 (H2A.X) residues which mark mitosis and gene repression, and DNA damage, respectively. Gene repression may also result from the sumoylation of H2A’s lysine 126 residue. In addition, ubiquitination of the H2A lysine 119 residue may play a role in spermatogenesis. It is the pattern of different modifications on several histone residues that alter chromatin, rather than modification of single residues.

Immunogen

KLH-conjugated linear peptide corresponding to the C-terminus of human Histone H2A phosphorylated at Thr120.

Application

Detect Histone H2A using this rabbit polyclonal antibody, Anti-phospho Histone H2A Antibody (Thr120) validated for use in western blotting, Dot Blot & IP.
Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated phospho Histone H2A (Thr120) in 500 µg of colcemid treated HeLa acid extract.

Dot Blot (Specificity) Analysis: 0.5 µg/mL from a representative lot detected phospho Histone H2A (Thr120) and not non-phosphorylated Histone H2A peptide or other peptides corresponding to modified histones.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Evaluated by Western Blotting in untreated and colcemid treated HeLa acid extract.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected phospho Histone H2A (Thr120) in 10 µg of colcemid treated HeLa acid extract, and demonstrated a decrease in signal in untreated HeLa acid extract.

Target description

~16 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service