Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE2627

Sigma-Aldrich

Anti-BMAL1

from rabbit

Synonym(s):

Aryl hydrocarbon receptor nuclear translocator-like protein 1, Basic-helix-loop-helix-PAS protein MOP3, Brain and muscle ARNT-like 1, Class E basic helix-loop-helix protein 5, bHLHe5, Member of PAS protein 3, PAS domain-containing protein 3, bHLH-PAS pro

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ARNTL(406)
mouse ... Arntl(11865)
rat ... Arntl(29657)

General description

Aryl hydrocarbon receptor nuclear translocator-like protein 1 (UniProt: O00327 also known as Basic-helix-loop-helix-PAS protein MOP3, Brain and muscle ARNT-like 1, BMAL1, Class E basic helix-loop-helix protein 5, bHLHe5; Member of PAS protein 3, PAS domain-containing protein 3, bHLH-PAS protein JAP3) is encoded by the ARNTL (also known as BHLHE5; BMAL1; MOP3; PASD3) gene (gene ID 406) in human. BMLA1 is highly expressed in the adult brain, skeletal muscle, and heart. It is a transcriptional activator that forms a core component of the circadian clock. These components include ARNTL/BMAL1, CLOCK, NPAS2, ARNTL2/BMAL2, PER1, PER2, PER3, CRY1, and CRY2. CLOCK or NPAS2 and ARNTL/BMAL1 or ARNTL2/BMAL2 form the positive limb of the feedback loop. They act in the form of a heterodimer and activate the transcription of core clock genes and clock-controlled genes that are involved in key metabolic processes. PER1/2/3 and CRY1/2, act as transcriptional repressors and form the negative limb of the feedback loop. They interact with the CLOCK|NPAS2-ARNTL/BMAL1|ARNTL2/BMAL2 heterodimer inhibiting its activity. BMLA1 is reported to be phosphorylated upon dimerization with CLOCK and this phosphorylation enhances its transcriptional activity and reduces the stability of the CLOCK-ARNTL/BMAL1 heterodimer by promoting its degradation. BMAL1 is shown to positively regulate myogenesis and negatively regulate adipogenesis via the transcriptional control of the genes of the canonical Wnt signaling pathway. BMAL1 is the only component of the mammalian circadian clock whose deletion in a murine model is shown to generate arrhythmicity. BMAL1 null-mice are reported to have reproductive problems, reduced locomotor activity, and they age quickly.

Specificity

Detects BMAL1 (Aryl hydrocarbon receptor nuclear translocator-like protein 1) in HeLa cell nuclear extract.

Immunogen

Epitope: C-terminus
KLH-conjugated peptide corresponding to the C-terminal 20 amino acids of human BMAL1 protein.

Application

Anti-BMAL1, Cat. No. ABE2627, is a highly specific rabbit polyclonal antibody that targets BMAL1 (Aryl hydrocarbon receptor nuclear translocator-like protein 1) and has been validated for use in Immunoprecipitation and Western Blotting
Immunoprecipitation Analysis: A representative lot immunoprecipitated BMAL1 in U2OS cells (Michael, A.K., et al. (2015). Mol Cell. 58(5):743-754).

Western Blotting Analysis: A representative lot detected BMAL1 protein expression in circadian synchronized NCI-H1299 cells (Michael, A.K., et al. (2015). Mol Cell. 58(5):743-754).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HeLa nuclear extract.

Western Blotting Analysis: 1 µg/mL of this antibody detected BMAL1 in 10 µg of HeLa nuclear extract.

Target description

~70 kDa observed. 68.8 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity purified.
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service