Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE1948

Sigma-Aldrich

Anti-CLIM-1/2 Antibody

serum, from rabbit

Synonym(s):

LIM domain-binding protein 2, LDB-2, Carboxyl-terminal LIM domain-binding protein 1, CLIM-1, LIM domain-binding factor CLIM1, CLIM-1/2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$333.00

$333.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$333.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$333.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

chicken, hamster, human, mouse

technique(s)

ChIP: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LDB2(9079)

General description

LIM domain-binding protein 1 (UniProt P70662; CLIM-2, LDB-1) encoded by the murine Ldb2 gene (Clim2, Nli; Gene ID 16825) and LIM domain-binding protein 2 (UniProt O55203; CLIM-1, LDB-2, CLP-36) encoded by the murine Ldb2 gene (Clim1; Gene ID 16826) are LIM domain-interacting cofactors whose interaction with LIM homeodomain (LIM-HD) transcription factors are crucial for the development of specific types of interneurons and motor neurons during neural tube development. The ubiquitin ligase RLIM targets CLIM for proteasomal degradation, thereby inhibiting developmental LIM-HD activity.

Specificity

Reacts with both CLIM1 and CLIM2. Expected to react with all spliced isoforms of CLIM1 (CLIM-1a, CLIM-1b, and isoform 3) & CLIM2 (Visvader-a, Tran-a, Tran-b).

Immunogen

Recombinant protein corresponding to mouse CLIM-1.

Application

This Anti-CLIM-1/2 Antibody is validated for use in Western Blotting, Immunocytochemistry, Chromatin Immunoprecipitation (ChIP), Immunohistochemistry, Immunofluorescence for the detection of CLIM-1/2.
Western Blotting Analysis: A representative lots detected endogenous CLIM1/2 in CHO and murine pituitary gonadotroph Alpha T3 cells, as well as bacterially expressed and in vitro tranlsated recombinant CLIM1/2 (Ostendorff, H.P., et al. (2006). Dev Dyn. 235(3):786-791; Ostendorff, H.P., et al. (2002). Nature. 416(6876):99-103).
Immunocytochemistry Analysis: A representative lot revealed an inverse correlation between the expression level of endogenous CLIM1/2 and that of its negative regulator RLIM in HEK293, CHO, murine pituitary gonadotroph Alpha T3, and chicken ectodermal cells by dual fluorescent immunocytochemistry (Ostendorff, H.P., et al. (2002). Nature. 416(6876):99-103).
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected CLIM1/2 occupancy at the alphaGSU promoter in murine pituitary gonadotroph Alpha T3 cells (Ostendorff, H.P., et al. (2002). Nature. 416(6876):99-103).
Immunohistochemistry Analysis: A representative lot detected similar stage-dependent expression patterns between CLIM1/2 and the ubiquitin ligase RLIM in the developing neural tube during early mouse embryogenesis (Ostendorff, H.P., et al. (2006). Dev Dyn. 235(3):786-791).
Immunofluorescence Analysis: A representative lot revealed highly co-localized expressions of CLIM1/2 and RLIM on thoracic sections from E12.5 mouse embryos by dual fluorescent immunohistochemistry (Ostendorff, H.P., et al. (2006). Dev Dyn. 235(3):786-791).

Quality

Evaluated by Western Blotting in E16 embryonic mouse brain tissue lysate.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected CLIM-1/2 in 10 µg of E16 embryonic mouse brain tissue lysate.

Target description

~50/43 kDa observed. Uncharacterized band(s) may appear in some lysates.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service