Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE1944

Sigma-Aldrich

Anti-Aprataxin Antibody

serum, from rabbit

Synonym(s):

Aprataxin, FHA-HIT, Forkhead-associated domain histidine triad-like protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$322.00

$322.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$322.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$322.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

porcine (based on high sequence homology), bovine (based on high sequence homology), rat (based on high sequence homology), mouse (based on high sequence homology), nonhuman primates (based on high sequence homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... APTX(54840)

General description

Aprataxin (UniProt Q7Z2E3; also known as FHA-HIT, Forkhead-associated domain histidine triad-like protein) is encoded by the APTX (also known as AOA, AOA1, AXA1, EAOH, EOAHA) gene (Gene ID 54840) in human. Aprataxin contains an N-terminal forkhead-associated (FHA; a.a. 38-87) domain akin to that found in the DNA end damage repair factor polynucleotide kinase phosphatase (PNKP), followed by a histidine triad (HIT; a.a. 182-287) domain found in nucleoside hydrolases and transferases, and a C-terminal C2H2 type Zinc finger domain. Aprataxin exhibits both DNA- and RNA-binding activity and plays a role in reversing toxic 5′-adenylated (5′-AMP) DNA lesions generated by DNA ligases when encountering nicked DNA structures with abnormal DNA termini. Aprataxin functions as a DNA ligase proofreader to directly reverse damaged 5′-adenylated termini of DNA strand breaks that have been subjected to DNA damage-induced abortive processing by DNA ligases. Ataxia with oculomotor apraxia type 1 (AOA1) is an autosomal recessive neurological syndrome caused by an impairment in DNA repair as a result of APTX gene mutations. The related type 2 disorder AOA2 is caused by a Senataxin-deficiency due to SETX gene mutations.

Specificity

Expected to react with all 13 human aprataxin spliced isoforms reported by UniProt (Q7Z2E3-1 through Q7Z2E3-13).

Immunogen

Recombinant full-length human aprataxin.

Application

Anti-Aprataxin Antibody is an antibody against Aprataxin for use in Western Blotting, Immunoprecipitation.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Western Blotting Analysis: A representative lot detected wild-type and the V263G mutant form of recombinant human aprataxin (Kijas, A.W., et al. (2006). J. Biol. Chem. 281(20):13939-13948).
Western Blotting Analysis: A representative lot detected aprataxin in lysates from HeLa and control lymphoblastoid cell lines (C3ABR and C2ABR), but not from AOA1 cell lines (L939, L938) derived from patients with the neurological disorder ataxia with oculomotor apraxia type 1 (AOA1) (Becherel, O.J., et al. (2006). Hum. Mol. Genet. 15(14):2239-2249).
Immunoprecipitation Analysis: A representative lot co-immunoprecipitated nucleolar proteins nucleolin and nucleophosmin with aprataxin from C3ABR lymphoblastoid cell lysate (Becherel, O.J., et al. (2006). Hum. Mol. Genet. 15(14):2239-2249).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Aprataxin in 10 µg of HeLa cell lysate.

Target description

~43 kDa observed.

Physical form

Rabbit polyclonal antibody in serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service