Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

ABE1846

Sigma-Aldrich

Anti-phospho CENP-T (Ser47) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Centromere protein T, phospho CENP-T (Ser47), CENP-T, Interphase centromere complex protein 22

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

nonhuman primates (based on 100% sequence homology)

technique(s)

dot blot: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer47 )

Gene Information

human ... CENPT(80152)

General description

Centromere protein T (UniProt Q96BT3; also known as CENP-T, Interphase centromere complex protein 22) is encoded by the CENPT (also known as C16orf56, ICEN22) gene (Gene ID 80152) in human. Kinetochore complex-mediated chromosome interaction with microtubules from the mitotic spindle is essential for proper chromosome segregation during mitosis. The kinetochore is a macromolecular structure composed of almost 100 different proteins assembled upon the centromere in a cell cycle-regulated manner. CENP-T and 15 other chromatin proximal proteins constitute the Constitutive Centromere Associated Network (CCAN) present at centromeres throughout the cell cycle. Some CCAN proteins (e.g. CENPT/W complex and CENP-C) mediate DNA binding, while others (e.g. CENP-N) interact directly with CENP-A-containing nucleosomes. Evidence suggests that CENP-C and the CENP-T/W complex function as core kinetochore assembly factors in vertebrate cells.

Specificity

Specifically reacts with the immunogen peptide, but not the corresponding non-phosphorylated sequence, by dot blot analysis.

Immunogen

Epitope: CENP-T pSer47
Linear peptide corresponding to a human CENP-T sequence containing phosphorylated Ser47.

Application

Anti-phospho CENP-T (Ser47) Antibody is an antibody against phospho CENP-T for use in Immunocytochemistry, Dot Blot, Western Blotting.
Immunocytochemistry Analysis: 4.0 µg/mL from a representative lot detected phospho CENP-T (Ser47) in HUVEC and A431 cells.
Dot Blot Analysis: 1.0 µg/mL from a representative lot detected the immunogen peptide, but not the corresponding non-phosphorylated sequence, by dot blot analysis.
Immunocytochemistry Analysis: A representative lot detected a closely synchronized CENP-T pSer47 phosphorylation and Hec1 immunoreactivity at the centromeres throughout HeLa cell cycle, starting at Telo/G1 and peaking at metaphase, followed by a steady deline of ~80% toward anaphase (Gascoigne, K.E., et al. (2011). Cell. 145(3):410-422).
Western Blotting Analysis: A representative lot detected the in vitro phosphorylation of recombinant CENP-T on Ser47 by recombinant CDK2/cyclin A (Gascoigne, K.E., et al. (2011). Cell. 145(3):410-422).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: 4.0 µg/mL of this antibody detected phospho CENP-T (Ser47) in HeLa cells.

Target description

60.4 kDa calculated.

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing PBS with 0.05% sodium azide and 30% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinhong Song et al.
Nature communications, 12(1), 7010-7010 (2021-12-03)
Defects in chromosome-microtubule attachment can cause chromosomal instability (CIN), frequently associated with infertility and aggressive cancers. Chromosome-microtubule attachment is mediated by a large macromolecular structure, the kinetochore. Sister kinetochores of each chromosome are pulled by microtubules from opposing spindle-poles, a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service