Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE1798

Sigma-Aldrich

Anti-SMUG1

from rabbit, purified by affinity chromatography

Synonym(s):

Single-strand selective monofunctional uracil DNA glycosylase, hSMUG1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$414.00

$414.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$414.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$414.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (based on high sequence homology), mouse (based on high sequence homology), rhesus macaque (based on high sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SMUG1(23583)

General description

Single-strand selective monofunctional uracil DNA glycosylase (UniProt: Q53HV7; also known as SMUG1, hSMUG1) is encoded by the SMUG1 gene (Gene ID: 23583) in human. SMUG1 is a base excision repair enzyme that removes uracil and oxidized pyrimidines from DNA. It recognizes base lesions in the genome and initiates base excision DNA repair. It displays greater activity toward mismatches (U/G) compared to matches (U/A). SMUG1 excises uracil (U), 5-formyluracil (fU) and uracil derivatives bearing an oxidized group at C5 [5-hydroxyuracil (hoU) and 5-hydroxymethyluracil (hmU)] in ssDNA and dsDNA, but not analogous cytosine derivatives (5-hydroxycytosine and 5-formylcytosine), nor other oxidized bases. Human and rat SMUG1 exhibit about 84% sequence homology and mouse SMUG1 has 94% homology with rat SMUG1. SMUG1 was originally identified as a monofunctional uracil-DNA glycosylase selective for ssDNA, however, later it was shown to be active for U in dsDNA. SMUG1 displays the following substrate preference: ssDNA > dsDNA (G pair) = dsDNA (A pair) at low salt concentration; and dsDNA (G pair) > dsDNA (A pair) > ssDNA at high salt concentration. SMUG1 preferentially binds to a dsDNA substrate over an ssDNA substrate when both are present. SMUG1 is shown to be a pseudouridine synthase dyskerin (DKC1) partner that contributes to rRNA quality control, partly by regulating 5-hydroxymethyluridine levels. (Ref: Matsubara, M et al. (2003). Biochemistry 42(17): 4993-5002; Masaoka A., et al. (2003). Biochemistry. 42(17): 5003-5012; Jobert, L et al. (2013). Mol. Cell 49(2): 339-345).

Specificity

This polyclonal antibody detects SMUG1 in HeLa cell nuclear extract. It targets an epitope within 13 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 13 amino acids from the N-terminal region of human SMUG1.

Application

Anti-SMUG1, Cat. No. ABE1798, is a highly specific rabbit polyclonal antibody that targets single-strand selective monofunctional uracil DNA glycosylase and has been tested in Western Blotting.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HeLa cell nuclear extract.

Western Blotting Analysis: 2 µg/mL of this antibody detected SMUG1 in 10 µg of HeLa cell nuclear extract.

Target description

~34 kDa observed; 29.86 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service