Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABE1793

Sigma-Aldrich

Anti-TEX14

from rabbit, purified by affinity chromatography

Synonym(s):

Inactive serine/threonine-protein kinase TEX14, Protein kinase-like protein SgK307, Sugen kinase 307, Testis-expressed sequence 14, Testis-expressed sequence 14 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$401.00

$401.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$401.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$401.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... TEX14(56155)

General description

Inactive serine/threonine-protein kinase TEX14 (UniProt: Q8IWB6; also known as Protein kinase-like protein SgK307, Sugen kinase 307, Testis-expressed sequence 14, Testis-expressed sequence 14 protein) is encoded by the TEX14 (also known as SGK307) gene (Gene ID: 56155) in human. TEX14 is a testis expressed, Plk-1 regulated protein that is required both for the formation of intercellular bridges during meiosis and for kinetochore-microtubule attachment during mitosis. Intercellular bridges connect differentiating germ cells and are required for spermatogenesis and male fertility. Three isoforms of TEX14 have been described that are produced by alternative splicing. TEX14 promotes the conversion of midbodies into intercellular bridges via its interaction with CEP55. Its interaction with CEP55 inhibits the interaction between CEP55 and PDCD6IP/ALIX and TSG101, which blocks cell abscission and transforms midbodies into intercellular bridges. TEX14 has three ANK domains (aa 27-54; 55-84, and 88-117) and a protein kinase domain. The protein kinase domain is catalytically inactive. TEX14 is phosphorylated on threonine residues by CDK1 during early phases of mitosis, which promotes the interaction with PLK1 and recruitment to kinetochores. It is also phosphorylated on Ser437 by PLK1 during late prometaphase, which promotes the rapid depletion from kinetochores and its subsequent degradation by the APC/C complex. (Ref.: Mondal, G., et al. (2012). Mol. Cell 45(5); 680-695).

Specificity

This rabbit polyclonal antibody detects Inactive serine/threonine-protein kinase TEX14 in human cells. It targets an epitope within 12 amino acids from the C-terminal half. Immunogen sequence is conserved in all three isoforms.

Immunogen

KLH-conjugated linenar peptide corresponding to 12 amino acids from the C-terminal half of human Inactive serine/threonine-protein kinase TEX14.

Application

Anti-TEX14, Cat. No. ABE1793, is a highly specific rabbit polyclonal antibody that targets Inactive serine/threonine-protein kinase TEX14 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250-1,000 dilution from a representative lot detected TEX14 in human testis and human ovary tissues.

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected TEX14 in 10 µg of HepG2 cell lysate.

Target description

~150 kDa observed; 167.90 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service