Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE1472

Sigma-Aldrich

Anti-phospho eIF4B Antibody (Ser422)

from rabbit, purified by affinity chromatography

Synonym(s):

Eukaryotic translation initiation factor 4B, eIF-4B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (based on 100% sequence homology), bovine (based on 100% sequence homology), zebrafish (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer422)

Gene Information

human ... EIF4B(1975)

General description

Eukaryotic translation initiation factor 4B (eIF-4B), encoded by the gene name ELF4B, is an RNA-binding protein that is involved in translation of RNA molecules and consequent protein synthesis. ELF4B is required for the binding of mRNA to ribosomes. ELF4B also functions in close association with EIF4-F and EIF4-A. ELF4B has been demonstrated to bind near the 5′-terminal cap of mRNA in presence of EIF-4F and ATP. ELF4B promotes the ATPase activity and the ATP-dependent RNA unwinding activity of both EIF4-A and EIF4-F. ELF4B contains multiple domains, including an N-terminal RNA recognition motif (RRM) and a C-terminal Arginine Rich Motif (ARM) that facilitate RNA binding. eIF4B may play a specific role in the assembly of the 48S initiation complex. Its translational activity is enhanced by phosphorylation of its ser406 and ser422 residues via the Ras-MAPK and PI3-K/mTOR pathways. eIF4B specially targets RNA molecules involved in cell proliferation and survival in response to signals converging from these pathways.

Immunogen

KLH-conjugated linear peptide corresponding to human phospho eIF4B (Ser422) near the C-terminus.

Application

Anti-phospho eIF4B Antibody (Ser422) | ABE1472 is an antibody against phospho eIF4B Antibody for use in Western Blotting.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected phospho eIF4B (Ser422) in 10 µg of HeLa cell lysate.

Target description

~75 kDa observed

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service