Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

ABE1464

Sigma-Aldrich

Anti-monomethyl Histone H2B (Pro1)

from rabbit

Synonym(s):

Histone H2B type 1-C/E/F/G/I, Histone H2B.1 A, Histone H2B.a, H2B/a, Histone H2B.g, H2B/g, Histone H2B.h, H2B/h, Histone H2B.k, H2B/k, Histone H2B.l, H2B/l

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$182.00
100 μG
$498.00

$182.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$182.00
100 μG
$498.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

$182.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

inhibition assay: suitable (peptide)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

monomethylation (Pro1)

Gene Information

General description

Histone H2B type 1-C/E/F/G/I (UniProt: P62807; also known as Histone H2B.1 A, Histone H2B.a, H2B/a, Histone H2B.g, H2B/g, Histone H2B.h, H2B/h, Histone H2B.k, H2B/k, Histone H2B.l, H2B/l) is encoded by the HIST1H2BC (also known as H2BFL, HIST1H2BE, H2BFH, HIST1H2BF, H2BFG, HIST1H2BG, H2BFA, HIST1H2BI, H2BFK) gene (Gene ID: 3017, 9339, 8343, 8344, 8346, 8347) in human. Histones are basic nuclear proteins that are responsible for the nucleosome structure of chromatin in eukaryotes. They undergo a number of post-translational modifications (PTM) at their N- and C-terminal tails which can influence chromatin structure and stability; and consequently, transcription, and DNA replication and repair. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which DNA is wrapped in repeating units, called nucleosomes, which limits DNA accessibility to the cellular machineries, which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. Histone H2B is one of the main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail, H2B is involved with the structure of the nucleosomes of the ′beads on a string′ structure. Histone H2B can be monoubiquitinated on Lysine123 by RAD6/UBC2-BRE1 complex to form H2BK123Ub1 complex that gives a specific tag for epigenetic transcriptional activation and is also a prerequisite for H3K4me and H3K79me formation. H2BK123ub1 is reported to modulates the formation of double-strand breaks during meiosis and is a required for DNA-damage checkpoint activation. Methylation of N-terminus Proline 1 of histone H2B (Pme) by NTMT (CG1675) is one of the rare modification that has been addressed. Heat shock or arsenite treatment is shown to increase methylation on proline 1 in Drosophila cells. (Ref.: Villar-Garea, A., et al. (2012). Nucl. Acid Res. 40(4); 1536 1549; Desrosiers, R., and Tanguay, RM. (1998). J. Biol. Chem. 263(10):4686-4692).

Specificity

This rabbit polyclonal antibody specifically detect Histone H2B monomethylated on proline 1.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the first 9 amino acids (methiionine removed) from the N-terminus region of human Histone H2B1C with methylated proline.

Application

Anti-monomethyl Histone H2B (Pro1) Antibody, Cat. No. ABE1464, is a highly specific rabbit polyclonal antibody that targets Histone H2B monomethylated on proline 1 and has been tested for use in Western Blotting and Peptide Inhibition Assay.
Peptide Inhibition Analysis: A 1:500 dilution from a representative lot was used with HeLa acid extract for peptide block analysis.



Dot Blot Specificity Analysis: Unmodified and various modified Histone peptides were probed with Anti-monomethyl Histone H2B (Pro1) (20 ng (A), 10 ng (B), 5 ng (C), 2.5 ng (D) dilutions).



Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected monomethyl Histone H2B (Pro1) in HeLa, A431, HUVEC, and NIH3T3 cell lines.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HeLa cells acid extract.

Western Blotting Analysis: 12 µg/mL of this antibody detected monomethyl Histone H2B (Pro1) in HeLa acid extract

Target description

~16 kDa observed; 13.91 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service