Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABE1403

Sigma-Aldrich

Anti-Cdk1/cdc2 Antibody C-term

from rabbit, purified by affinity chromatography

Synonym(s):

Cyclin-dependent kinase 1, CDK1, Cell division control protein 2 homolog, Cell division protein kinase 1, p34 protein kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology), hamster (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

hamster ... Cdk1(100766716)
human ... CDK1(983)
mouse ... Cdk1(12534)
rat ... Cdk1(54237)

General description

Cyclin-dependent kinase 1 (Cdk1) is also known as Cell division control protein 2 homolog (Cdc2. ;Entry of all eukaryotic cells into mitosis is regulated by activation of cdc2 kinase. Activation of cdc2 is controlled at several steps including cyclin binding and phosphorylation of Thr161. However, the critical regulatory step in activating cdc2 during progression into mitosis appears to be dephosphorylation of Tyr15 and Thr14. Phosphorylation at Tyr15 and inhibition of cdc2 is carried out by Wee1 and Myt1 protein kinases, while Tyr15 dephosphorylation and activation of cdc2 is carried out by the cdc25 phosphatase. Expression of cdc2 plays a critical role in cell transition through the G2/M phase. The activity of cdc2 in dividing cells increases at the onset of M phase coincident with disassembly of the cell nucleus, generation of mitotic spindles and chromosome condensation. Several proteins including Histone H1 and pp60src are substrates for cdc2.

Immunogen

Epitope: Near C-terminus
KLH-conjugated linear peptide corresponding to human Cdk1/cdc2 C-term near the C-terminus.

Application

Immunoprecipitation Analysis: 2.0 µg/mL from a representative lot immunoprecipitated Cdk1/cdc2 in 500 µg of Balb/3T3/A31 cell lysate.
Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected Cdk1/cdc2 in human testis and human breast cancer tissue.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-Cdk1/cdc2 Antibody C-term is validated for use in Western Blotting and Immunohistochemistry and Immunoprecipitation for the detection of Cdk1/cdc2 C-term.

Quality

Evaluated by Western Blotting in Balb/3T3/A31 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected Cdk1/cdc2 in 10 µg of Balb/3T3/A31 cell lysate.

Target description

~34 kDa observed. Higher molecular weight band may correspond to phosphorylated form of protein.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiang-Wen Lee et al.
Molecules (Basel, Switzerland), 27(2) (2022-01-22)
Osteosarcoma, a primary bone tumor, responds poorly to chemotherapy and radiation therapy in children and young adults; hence, as the basis for an alternative treatment, this study investigated the cytotoxic and antiproliferative effects of naringenin on osteosarcoma cell lines, HOS
Rosemarie E Gough et al.
The Journal of biological chemistry, 297(1), 100837-100837 (2021-06-13)
Talin (TLN1) is a mechanosensitive component of adhesion complexes that directly couples integrins to the actin cytoskeleton. In response to force, talin undergoes switch-like behavior of its multiple rod domains that modulate interactions with its binding partners. Cyclin-dependent kinase-1 (CDK1)
Matthew C Jones et al.
The Journal of cell biology, 217(9), 3203-3218 (2018-06-23)
In most tissues, anchorage-dependent growth and cell cycle progression are dependent on cells engaging extracellular matrices (ECMs) via integrin-receptor adhesion complexes. In a highly conserved manner, cells disassemble adhesion complexes, round up, and retract from their surroundings before division, suggestive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service