Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABE1332

Sigma-Aldrich

Anti-monomethyl Histone H2A (Gln104)

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H2A type 1, Gln104 monomethylated, H2A.1, Gln104 monomethylated, Histone H2A/p, Gln104 monomethylated, H2AQ104me, Gln104 monomethylated

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$498.00

$498.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$498.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

$498.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

dot blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

monomethylation (Gln104)

Gene Information

General description

Histone H2A type 1 (UniProt P0C0S8; also known as H2A.1, Histone H2A/p) is encoded by the HIST1H2AG (also known as H2AFP, HIST1H2AI, H2AFC, HIST1H2AK, H2AFD, HIST1H2AL, H2AFI, HIST1H2AM, H2AFN) gene (Gene ID 8969) in human. Histone H2A is one of four components of the core nucleosome structure. The nucleosome represents a unit of chromatin in which DNA is wrapped around a histone octamer. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones. Histones undergo a number of post-translational modifications in response to various stimuli that may induce changes in the structure of the nucleosome and hide or expose DNA sequences. Histone H2A, in particular, undergoes acetylation on lysine 5 by Tip60 enzyme, which may promote unfolding of chromatin and transcription. In addition, H2A is phosphorylated on serine 1 and serine 139 (H2A.X) residues, which mark mitosis and gene repression and DNA damage, respectively. Gene repression may also result from the sumoylation of H2A s lysine 126 residue. In addition, ubiquitination of the H2A lysine 119 residue may play a role in spermatogenesis. Monoubiquitination of Lys-120 (H2AK119Ub) by RING1, TRIM27 and RNF2/RING2 complex gives a specific tag for epigenetic transcriptional repression and participates in X chromosome inactivation of female mammals.

Specificity

Specificity of this polyclonal antibody was verified by dot blot analysis. This polyclonal antibody specifically detected peptide corresponding to histone H2A target region sequence with monomethylated Gln104 (Q104me), but not H2A peptides without Q104me modification, nor peptides derived from histone H1, H2A.Z, H2B, H3, H3.3, or H4 sequences.

Immunogen

KLH-conjugated linear peptide corresponding to human histone H2A target region sequence containing monomethylated Gln104 (Q104me).

Application

Anti-monomethyl Histone H2A (Gln104) Antibody, Cat. No. ABE1332, is a highly specific rabbit polyclonal antibody that targets histone H2A Gln104 monomethylation (Q104me) and has been tested by histone peptide array Dot Blot.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Dot Blot Analysis of histone peptide array.

Dot Blot Analysis: A 1:200 dilution of this antibody detected 4-40 ng of histone H2A peptide containing monomethylated Gln104 (Q104me).

Target description

13.96 kDa (Met1 removed) calculated.

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service